Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SIRT2 Antibodies

          Rockland

          SIRT2 Polyclonal Antibody

          View all (22) SIRT2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SIRT2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          SIRT2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SIRT2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIRT2 Antibody (600-401-EL7) in ICC/IF

          Immunofluorescence Microscopy of SIRT2 antibody. Tissue: Human brain tissue. Fixation: 0.5% PFA. Antigen retrieval: not required. Primary antibody: SIRT2 antibody at 20 µg/mL for 1 hr at RT. Secondary antibody: Fluorescein rabbit secondary antibody at 1:10,000 for 45 min at RT. Staining: SIRT2 as a red fluorescent signal. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIRT2 Antibody in Immunocytochemistry (ICC/IF)
          SIRT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SIRT2 Antibody in Western Blot (WB)
          SIRT2 Polyclonal Antibody

          Product Details

          600-401-EL7

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2.5-5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2.5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          20 µg/mL
          -

          ELISA (ELISA)

          1:25,000-1:100,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Anti-SIRT2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 17 amino acid synthetic peptide near the N-terminus of the human SIRT2.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5E5 antigen; mSIR2L2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sirtuin-2; NAD-dependent protein deacylase sirtuin-2; NAD-dependent protein defatty-acylase sirtuin-2; Regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2-h; SIR2-like protein 2; sir2-related protein type 2; sir2-related protein type 2; sirtuin type 2; NAD-dependent deacetylase sirtuin-2; silent information regulator 2; SIR2-like protein 2; regulatory protein SIR2; sirtuin type 2; sirtuin-2; unnamed protein product; yeast silent information regulator 2 (Sir2p) homolog

          View more View less

          Gene Aliases: 5730427M03Rik; SIR2; SIR2L; SIR2L2; SIRT2

          View more View less

          UniProt ID: (Human) Q8IXJ6, (Mouse) Q8VDQ8

          View more View less

          Entrez Gene ID: (Human) 22933, (Mouse) 64383, (Rat) 361532

          View more View less

          Function(s)
          chromatin binding NAD+ ADP-ribosyltransferase activity histone deacetylase activity protein binding zinc ion binding transferase activity NAD-dependent histone deacetylase activity protein deacetylase activity NAD-dependent protein deacetylase activity histone acetyltransferase binding histone deacetylase binding tubulin deacetylase activity ubiquitin binding metal ion binding NAD-dependent histone deacetylase activity (H4-K16 specific) NAD binding NAD+ binding DNA-binding transcription factor binding NAD-dependent protein demyristoylase activity NAD-dependent protein depalmitoylase activity protein ADP-ribosylase activity transcription factor binding protein-N-terminal asparagine amidohydrolase activity UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity hydrolase activity hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in linear amides iprodione amidohydrolase activity (3,5-dichlorophenylurea)acetate amidohydrolase activity 4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity didemethylisoproturon amidohydrolase activity N-isopropylacetanilide amidohydrolase activity N-cyclohexylformamide amidohydrolase activity isonicotinic acid hydrazide hydrolase activity cis-aconitamide amidase activity gamma-N-formylaminovinylacetate hydrolase activity N2-acetyl-L-lysine deacetylase activity O-succinylbenzoate synthase activity indoleacetamide hydrolase activity N-acetylcitrulline deacetylase activity N-acetylgalactosamine-6-phosphate deacetylase activity beta-tubulin binding diacetylchitobiose deacetylase activity chitooligosaccharide deacetylase activity molecular_function
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated protein deacetylation autophagy cell cycle mitotic nuclear division nervous system development negative regulation of cell proliferation negative regulation of autophagy negative regulation of peptidyl-threonine phosphorylation phosphatidylinositol 3-kinase signaling histone deacetylation myelination in peripheral nervous system cell differentiation regulation of myelination positive regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to oxidative stress peptidyl-lysine deacetylation cellular response to hepatocyte growth factor stimulus negative regulation of protein catabolic process negative regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of DNA binding protein kinase B signaling cellular lipid catabolic process regulation of fat cell differentiation negative regulation of fat cell differentiation positive regulation of meiotic nuclear division negative regulation of striated muscle tissue development negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter hepatocyte growth factor receptor signaling pathway negative regulation of oligodendrocyte differentiation cell division meiotic cell cycle regulation of cell cycle positive regulation of cell division positive regulation of attachment of spindle microtubules to kinetochore negative regulation of transcription from RNA polymerase II promoter in response to hypoxia cellular response to caloric restriction negative regulation of oligodendrocyte progenitor proliferation histone H3 deacetylation histone H4 deacetylation cellular response to molecule of bacterial origin cellular response to hypoxia cellular response to epinephrine stimulus tubulin deacetylation positive regulation of execution phase of apoptosis positive regulation of oocyte maturation negative regulation of NLRP3 inflammasome complex assembly negative regulation of defense response to bacterium ripoptosome assembly involved in necroptotic process negative regulation of reactive oxygen species metabolic process positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia central nervous system development substantia nigra development chromatin silencing at rDNA immune system process chromatin organization ubiquitin-dependent protein catabolic process fatty acid biosynthetic process mitotic nuclear envelope reassembly regulation of exit from mitosis skeletal muscle satellite cell differentiation lipid catabolic process heterochromatin assembly telomeric heterochromatin assembly regulation of gene expression, epigenetic regulation of phosphorylation post-translational protein modification NLRP3 inflammasome complex assembly innate immune response negative regulation of fatty acid biosynthetic process positive regulation of fatty acid biosynthetic process regulation of lipid biosynthetic process negative regulation of developmental process regulation of multicellular organismal process response to redox state positive regulation of small molecule metabolic process negative regulation of satellite cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline