Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SIRT7 Antibodies

          Proteintech

          SIRT7 Polyclonal Antibody

          23 References
          View all (20) SIRT7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SIRT7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIRT7 Antibody (12994-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded mouse heart tissue slide using 12994-1-AP (SIRT7 antibody) at dilution of 1:50 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SIRT7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SIRT7 Antibody in Western Blot (WB)
          SIRT7 Antibody in Western Blot (WB)
          SIRT7 Antibody in Immunoprecipitation (IP)
          SIRT7 Polyclonal Antibody

          Product Details

          12994-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SIRT7 Fusion Protein Ag3656 (1-400 aa encoded by BC017305)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MAAGGLSRS ERKAAERVRR LREEQQRERL RQVSRILRKA AAERSAEEGR LLAESADLVT ELQGRSRRRE GLKRRQEEVC DDPEELRGKV RELASAVRNA KYLVVYTGAG ISTAASIPDY RGPNGVWTLL QKGRSVSAAD LSEAEPTLTH MSITRLHEQK LVQHVVSQNC DGLHLRSGLP RTAISELHGN MYIEVCTSCV PNREYVRVFD VTERTALHRH QTGRTCHKCG TQLRDTIVHF GERGTLGQPL NWEAATEAAS RADTILCLGS SLKVLKKYPR LWCMTKPPSR RPKLYIVNLQ WTPKDDWAAL KLHGKCDDVM RLLMAELGLE IPAYSRWQDP IFSLATPLRA GEEGSHSRKS LCRSREEAPP GDRGAPLSSA PILGGWFGRG CTKRTKRKKV T (1-400 aa encoded by BC017305)

          Target Information

          SIRT7 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. SIRT7 is included in class IV. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. SIRT7 is included in class IV of the sirtuin family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC126840; MGC126842; NAD-dependent deacetylase sirtuin-7; NAD-dependent protein deacetylase sirtuin-7; NAD-dependent protein deacylase sirtuin-7; Regulatory protein SIR2 homolog 7; silent mating type information regulation 2, S.cerevisiae, homolog 7; SIR2-like protein 7; sir2-related protein type 7; sirtuin type 7; sirtuin7; unnamed protein product

          View more View less

          Gene Aliases: SIR2L7; SIRT7

          View more View less

          UniProt ID: (Human) Q9NRC8, (Mouse) Q8BKJ9, (Rat) B2RZ55

          View more View less

          Entrez Gene ID: (Human) 51547, (Mouse) 209011, (Rat) 303745

          View more View less

          Function(s)
          chromatin binding protein binding protein methyltransferase activity transferase activity hydrolase activity NAD-dependent protein deacetylase activity protein-succinyllysine desuccinylase activity metal ion binding protein-glutaryllysine deglutarylase activity NAD+ binding NAD-dependent histone deacetylase activity (H3-K18 specific) NAD-dependent protein-lysine depropionylase activity histone deacetylase activity protein-N-terminal asparagine amidohydrolase activity UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity iprodione amidohydrolase activity (3,5-dichlorophenylurea)acetate amidohydrolase activity 4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity didemethylisoproturon amidohydrolase activity N-isopropylacetanilide amidohydrolase activity N-cyclohexylformamide amidohydrolase activity isonicotinic acid hydrazide hydrolase activity cis-aconitamide amidase activity gamma-N-formylaminovinylacetate hydrolase activity N2-acetyl-L-lysine deacetylase activity O-succinylbenzoate synthase activity indoleacetamide hydrolase activity N-acetylcitrulline deacetylase activity N-acetylgalactosamine-6-phosphate deacetylase activity diacetylchitobiose deacetylase activity chitooligosaccharide deacetylase activity molecular_function
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter osteoblast differentiation regulation of gluconeogenesis DNA repair regulation of DNA repair chromatin organization regulation of transcription from RNA polymerase II promoter protein deacetylation cellular response to DNA damage stimulus synapsis rRNA transcription negative regulation of transposition, RNA-mediated regulation of mitochondrion organization negative regulation of protein ubiquitination regulation of gene expression, epigenetic protein kinase B signaling positive regulation of gluconeogenesis negative regulation of gene expression, epigenetic positive regulation of gene expression, epigenetic positive regulation of transcription from RNA polymerase I promoter regulation of protein export from nucleus negative regulation of protein kinase B signaling protein deglutarylation R-loop processing protein depropionylation DNA repair-dependent chromatin remodeling regulation of transcription of nucleolar large rRNA by RNA polymerase I positive regulation of rRNA processing transcription, DNA-templated regulation of transcription, DNA-templated positive regulation of transcription involved in exit from mitosis chromatin modification histone H3 deacetylation histone H4 deacetylation biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0