Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SKIV2L Antibodies

          Abnova

          SKIV2L Polyclonal Antibody, MaxPab™

          View all (19) SKIV2L antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SKIV2L Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          SKIV2L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SKIV2L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SKIV2L Antibody (H00006499-B01P) in WB

          Western Blot analysis of SKIV2L expression in transfected 293T cell line by SKIV2L MaxPab polyclonal antibody. Lane 1: SKIV2L transfected lysate(137.06 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SKIV2L Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          H00006499-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SKIV2L (ABW03481.1, 1 a.a. approximately 1246 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MMDTERLVLP PPDPLDLPLR AVELGCTGHW ELLNLPGAPE SSLPHGLPPC APDLQQEAEQ LFLSSPAWLP LHGVEHSARK WQRKTDPWSL LAVLGAPVPS DLQAQRHPTT GQILGYKEVL LENTNLSATT SLSLRRPPGP ASQSLWGNPT RYPFWPGGMD EPTITDLNTR EEAEEEIDFE KDLLTIPPGF KKGMDFAPKD CPTPAPGLLS LSCLLEPLDL GGGDEDENEA VGQPGGPRGD TVSASPCSAP LARASSLEDL VLKEASTAVS TPEAPEPPSQ EQWAIPVDAT SPVGDFYRLI PQPAFQWAFE PDVFQKQAIL HLERHDSVFV AAHTSAGKTV VAEYAIALAQ KHMTRTIYTS PIKALSNQKF RDFRNTFGDV GLLTGDVQLH PEASCLIMTT EILRSMLYSG SDVIRDLEWV IFDEVHYIND VERGVVWEEV LIMLPDHVSI ILLSATVPNA LEFADWIGRL KRRQIYVIST VTRPVPLEHY LFTGNSSKTQ GELFLLLDSR GAFHTKGYYA AVEAKKERMS KHAQTFGAKQ PTHQGGPAQD RGVYLSLLAS LRTRAQLPVV VFTFSRGRCD EQASGLTSLD LTTSSEKSEI HLFLQRCLAR LRGSDRQLPQ VLHMSELLNR GLGVHHSGIL PILKEIVEML FSRGLVKVLF ATETFAMGVN MPARTVVFDS MRKHDGSTFR DLLPGEYVQM AGRAGRRGLD PTGTVILLCK GRVPEMADLH RMMMGKPSQL QSQFRLTYTM ILNLLRVDAL RVEDMMKRSF SEFPSRKDSK AHEQALAELT KRLGALEEPD MTGQLVDLPE YYSWGEELTE TQHMIQRRIM ESVNGLKSLS AGRVVVVKNQ EHHNALGVIL QVSSNSTSRV FTTLVLCDKP LSQDPQDRGP ATAEVPYPDD LVGFKLFLPE GPCDHTVVKL QPGDMAAITT KVLRVNGEKI LEDFSKRQQP KFKKDPPLAA VTTAVQELLR LAQAHPAGPP TLDPVNDLQL KDMSVVEGGL RARKLEELIQ GAQCVHSPRF PAQYLKLRER MQIQKEMERL RFLLSDQSLL LLPEYHQRVE VLRTLGYVDE VGTVKLAGRV ACAMSSHELL LTELMFDNAL STLRPEEIAA LLSGLVCQSP GDAGDQLPNT LKQGIERVRA VAKRIGEVQV ACGLNQTVEE FVGELNFGLV EVVYEWARGM PFSELAGLSG TPEGLVVRCI QRLAEMCRSL RGAARLVGEP VLGAKMETAA TLLRRDIVFA ASLYTQ

          Target Information

          DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: helicase SKI2W; helicase-like protein; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; SKI2 subunit of superkiller complex protein; superkiller viralicidic activity 2-like; unnamed protein product

          View more View less

          Gene Aliases: 170A; DDX13; HLP; SKI2; SKI2W; SKIV2; SKIV2L; SKIV2L1; THES2

          View more View less

          UniProt ID: (Human) Q15477

          View more View less

          Entrez Gene ID: (Human) 6499

          View more View less

          Function(s)
          nucleic acid binding RNA binding RNA helicase activity protein binding ATP binding ATPase activity 3'-5' RNA helicase activity
          Process(es)
          nuclear-transcribed mRNA catabolic process RNA catabolic process mRNA catabolic process nuclear-transcribed mRNA catabolic process, 3'-5' exonucleolytic nonsense-mediated decay nuclear-transcribed mRNA catabolic process, non-stop decay rescue of stalled ribosome
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-gw77j:80/100.66.76.64:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0