Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody specifically recognized the endogenous SLC25A1; no protein was detected in cells from subjects containing two alleles with truncating mutations with this antibody.
Immunogen sequence: MPAPRAPRA LAAAAPASGK AKLTHPGKAI LAGGLAGGIE ICITFPTEYV KTQLQLDERS HPPRYRGIGD CVRQTVRSHG VLGLYRGLSS LLYGSIPKAA VRFGMFEFLS NHMRDAQGRL DSTRGLLCGL GAGVAEAVVV VCPMETIKVK FIHDQTSPNP KYRGFFHGVR EIVREQGLKG TYQGLTATVL KQGSNQAIRF FVMTSLRNWY RGDNPNKPMN PLITGVFGAI AGAASVFGNT PLDVIKTRMQ GLEAHKYRNT WDCGLQILKK EGLKAFYKGT VPRLGRVCLD VAIVFVIYDE VVKLLNKVWK TD (1-311 aa encoded by BC004980)
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: citrate carrier; citrate isocitrate carrier; citrate transport protein; citrate transporter) member 1; DiGeorge syndrome gene j; mitochondrial citrate carrier; mitochondrial tricarboxylate carrier; pmCiC; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; solute carrier family 25 member 1; tricarboxylate carrier protein; txtp; unnamed protein product
Gene Aliases: 1300019P08Rik; 2610100G11Rik; CIC; CMS23; CTP; D2L2AD; Dgsj; SEA; SLC20A3
UniProt ID: (Human) P53007, (Rat) P32089, (Mouse) Q8JZU2
Entrez Gene ID: (Human) 6576, (Rat) 29743, (Mouse) 13358
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support