Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SLC26A7 Antibodies

          Proteintech

          SLC26A7 Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SLC26A7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SLC26A7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SLC26A7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SLC26A7 Antibody (17654-1-AP) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 17654-1-AP (SLC26A7 antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SLC26A7 Antibody in Western Blot (WB)
          SLC26A7 Antibody in Western Blot (WB)
          SLC26A7 Polyclonal Antibody

          Product Details

          17654-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SLC26A7 fusion protein Ag11901
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          37.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: IAIVIGRFPR AMTVSIKNMK EMEFKVKTEM DSETLQQVKI ISINNPLVFL NAKKFYTDLM NMIQKENACN QPLDDISKCE QNTLLNSLSN GNCNEEASQS CPNEKCYLIL DCSGFTFFDY SGVSMLVEVY MDCKGRSVDV LLAHCTASLI KAMTYYGNLD SEKPIFFESV SAAISHIHSN KNLSKLSDHS EV

          Target Information

          SLC26A7 (solute carrier family 26 member 7) is a chloride-base exchanger and/or chloride transporter that is expressed on basolateral membrane of acid-secreting cells in renal outer medullary collecting duct (OMCD) and in gastric parietal cells also. SLC26A7 belongs to conserved family of anion transporters SLC26 isoforms which display tissue-specific expression patterns in epithelial cells and several SLC26 members function as chloride/bicarbonate exchangers that include SLC26A3 (DRA), SLC26A4 (pendrin), SLC26A6 (PAT1 or CFEX), SLC26A7, and SLC26A9 among which SLC26A7 and SLC26A9 also function as chloride channels. In renal tissues, Slc26a7 co-localizes with AE1, a well-known Cl-/HCO3- exchanger, on the basolateral membrane of (acid-secreting) A-intercalated cells in OMCD cells, and in the gastric tissue, SLC26A7 co-localizes with AE2, a major Cl-/HCO3- exchanger, on the basolateral membrane of acid secreting parietal cells. SLC26A7 acts as a sodium-independent DIDS-sensitive anion exchanger mediating bicarbonate, chloride, sulfate and oxalate transport. SLC26A7 maintains the electrolyte/acid-base homeostasis in kidneys by acting as a distal excretory segment-specific anion exchanger, specifically chloride, and plays a major role in gastric acid secretion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Anion exchange transporter; anion exchanger Slc26a7; member of the SLC26 family; MGC30955; RP23-272I16.1; slc26a7; solute carrier family 26 (anion exchanger), member 7; Solute carrier family 26 member 7; solute carrier family 26, member 7; solute carrier family 6 member 7; sulfate anion transporter; sulfate anion transporter 2

          View more View less

          Gene Aliases: SLC26A7; SUT2

          View more View less

          UniProt ID: (Human) Q8TE54, (Mouse) Q8R2Z3

          View more View less

          Entrez Gene ID: (Human) 115111, (Rat) 297910, (Mouse) 208890

          View more View less

          Function(s)
          anion channel activity chloride channel activity bicarbonate transmembrane transporter activity sulfate transmembrane transporter activity antiporter activity oxalate transmembrane transporter activity chloride:bicarbonate antiporter activity secondary active sulfate transmembrane transporter activity anion transmembrane transporter activity anion:anion antiporter activity transporter
          Process(es)
          gastric acid secretion thyroid hormone generation ion transport anion transport chloride transport bicarbonate transport iodide transport nitrate transport oxalate transport gluconate transmembrane transport transmembrane transport sulfate transmembrane transport chloride transmembrane transport carboxylic acid transmembrane transport sulfate transport inorganic anion transport ion transmembrane transport transport anion transmembrane transport organic acid transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline