Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SLC38A3 Antibodies

          Proteintech

          SLC38A3 Polyclonal Antibody

          21 References
          View all (6) SLC38A3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SLC38A3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          SLC38A3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SLC38A3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SLC38A3 Antibody (14315-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol) fixed HEK-293 cells using 14315-1-AP (SLC38A3 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SLC38A3 Antibody in Immunocytochemistry (ICC/IF)
          SLC38A3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SLC38A3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Flow Cytometry (Flow)
          SLC38A3 Antibody in Immunoprecipitation (IP)
          SLC38A3 Polyclonal Antibody

          Product Details

          14315-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SLC38A3 Fusion Protein Ag5553 (1-191 aa encoded by BC042875)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MEAPLQTEM VELVPNGKHS EGLLPVITPM AGNQRVEDPA RSCMEGKSFL QKSPSKEPHF TDFEGKTSFG MSVFNLSNAI MGSGILGLAY AMANTGIILF LFLLTAVALL SSYSIHLLLK SSGVVGIRAY EQLGYRAFGT PGKLAAALAI TLQNIGAMSS YLYIIKSELP LVIQTFLNLE EKTSDWYMNG NY (1-191 aa encoded by BC042875)

          Target Information

          As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hSN1; mNAT; N system amino acids transporter NAT-1; N-system amino acid transporter 1; Na(+)-coupled neutral amino acid transporter 3; SLC38A3; slc38a3 {ECO:0000312|RGD:628620}; Sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; System N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter; unnamed protein product

          View more View less

          Gene Aliases: 0610012J02Rik; D9Ucla2; DEE102; G17; mNAT; NAT1; Slc38-3; SLC38A3; SN1; SNAT3

          View more View less

          UniProt ID: (Human) Q99624, (Rat) Q9JHZ9, (Mouse) Q9DCP2

          View more View less

          Entrez Gene ID: (Human) 10991, (Rat) 252919, (Mouse) 76257

          View more View less

          Function(s)
          L-histidine transmembrane transporter activity neutral amino acid:sodium symporter activity amino acid transmembrane transporter activity L-alanine transmembrane transporter activity L-asparagine transmembrane transporter activity L-glutamine transmembrane transporter activity symporter activity antiporter activity L-glutamine, sodium:proton antiporter activity L-asparagine, sodium:proton antiporter activity L-histidine, sodium:proton antiporter activity amino acid transporter
          Process(es)
          amino acid transmembrane transport ion transport sodium ion transport amino acid transport asparagine transport glutamine transport female pregnancy glutamine secretion L-alanine transport histidine transport positive regulation of transcription from RNA polymerase II promoter cellular response to potassium ion starvation amino acid homeostasis transport across blood-brain barrier L-histidine transport L-glutamine import across plasma membrane L-histidine import across plasma membrane L-asparagine import across plasma membrane positive regulation of glutamine transport positive regulation of transcription from RNA polymerase II promoter in response to acidic pH L-histidine transmembrane transport transport drug transmembrane transport cation transmembrane transport L-alpha-amino acid transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline