Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: IPDTVTQYS ELYHDFTTTQ VGMHDIVRSF RQTMDQYSTE PGRYRFMGTE AYAESIDRTV MYYGLPFIQE ADFPFNNYLS MLDTVSGNSV YEVITSWMEN MPEGKWPNWM IGGPDSSRLT SRLGNQYVNV MNMLLFTLPG TPITYYGEEI GMGNIVAANL NESYDINTLR SKSPMQWDNS SNAGFSEASN TWLPTNSDYH TVNVDVQKTQ PRSALKLYQD LSLLHANELL LNRGWFCHLR NDSHYVVYTR ELDGIDRIFI VVLNFGESTL LNLHNMISGL PAKIRIRLST NSADKGSKVD TSGIFLDKGE GLIFEHNTKN LLHRQTAFRD RCFVSNRACY SSVLNILYTS C (336-685 aa encoded by BC022386)
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amino acid transporter 1; B(0,+)-type amino acid transport protein; b(0,+)-type amino acid transporter-related heavy chain; D2; FLJ34681; NAA-TR; neutral and basic amino acid transport protein rBAT; neutral and basic amino acid transporter protein; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine dibasic and neutral amino acid transporters activator of cystine dibasic and neutral amino acid transport) member 1 isoform SLC3A1_1; solute carrier family 3 (cystine dibasic and neutral amino acid transporters activator of cystine dibasic and neutral amino acid transport) member 1 isoform SLC3A1_2; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; unnamed protein product
Gene Aliases: ATR1; CSNU1; D2; D2H; NBAT; NTAA; RBAT
UniProt ID: (Human) Q07837, (Rat) Q64319, (Mouse) Q91WV7
Entrez Gene ID: (Human) 6519, (Rat) 29484, (Mouse) 20532
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support