Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: YLFSSFTIDL PWGGCYHEWN TEHCMEFQKT NGSLNGTSEN AT
Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 83%.
Slc6a13 catalyzes sodium and chloride dependent transport of gamma-aminobutyric acid (GABA) to terminate GABA mediated synaptic activity. Slc6a13 is also a high-affinity gamma-aminobutyric acid (GABA) transporter, terminates biological actions of GABA by reuptake into cell, and its reuptake function utilizes sodium and chloride concentration gradients. Further, Slc6a13 may also aid in beta-alanine transport. Diseases associated with SLC6A13 include Traumatic Glaucoma and Neovascular Glaucoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support