Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SMPD3 Antibodies

          Abnova

          SMPD3 Polyclonal Antibody, MaxPab™

          View all (10) SMPD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SMPD3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SMPD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SMPD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SMPD3 Antibody (H00055512-B01P) in WB

          Western Blot analysis of SMPD3 expression in transfected 293T cell line (H00055512-T02) by SMPD3 MaxPab polyclonal antibody.

          Lane 1: SMPD3 transfected lysate(72.05 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SMPD3 Antibody in Western Blot (WB)
          SMPD3 Polyclonal Antibody, MaxPab™

          Product Details

          H00055512-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SMPD3 (NP_061137.1, 1 a.a. approximately 655 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MVLYTTPFPN SCLSALHCVS WALIFPCYWL VDRLAASFIP TTYEKRQRAD DPCCLQLLCT ALFTPIYLAL LVASLPFAFL GFLFWSPLQS ARRPYIYSRL EDKGLAGGAA LLSEWKGTGP GKSFCFATAN VCLLPDSLAR VNNLFNTQAR AKEIGQRIRN GAARPQIKIY IDSPTNTSIS AASFSSLVSP QGGDGVARAV PGSIKRTASV EYKGDGGRHP GDEAANGPAS GDPVDSSSPE DACIVRIGGE EGGRPPEADD PVPGGQARNG AGGGPRGQTP NHNQQDGDSG SLGSPSASRE SLVKGRAGPD TSASGEPGAN SKLLYKASVV KKAAARRRRH PDEAFDHEVS AFFPANLDFL CLQEVFDKRA ATKLKEQLHG YFEYILYDVG VYGCQGCCSF KCLNSGLLFA SRYPIMDVAY HCYPNKCNDD ALASKGALFL KVQVGSTPQD QRIVGYIACT HLHAPQEDSA IRCGQLDLLQ DWLADFRKST SSSSAANPEE LVAFDVVCGD FNFDNCSSDD KLEQQHSLFT HYRDPCRLGP GEEKPWAIGT LLDTNGLYDE DVCTPDNLQK VLESEEGRRE YLAFPTSKSS GQKGRKELLK GNGRRIDYML HAEEGLCPDW KAEVEEFSFI TQLSGLTDHL PVAMRLMVSS GEEEA

          Target Information

          Catalyzes the hydrolysis of sphingomyelin to form ceramide and phosphocholine. Ceramide mediates numerous cellular functions, such as apoptosis and growth arrest, and is capable of regulating these 2 cellular events independently. Also hydrolyzes sphingosylphosphocholine. Regulates the cell cycle by acting as a growth suppressor in confluent cells. Probably acts as a regulator of postnatal development and participates in bone and dentin mineralization.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Neutral sphingomyelinase 2; Neutral sphingomyelinase II; nSMase-2; SMPD3; Sphingomyelin phosphodiesterase 3; unnamed protein product

          View more View less

          Gene Aliases: NSMASE2; SMPD3

          View more View less

          UniProt ID: (Human) Q9NY59

          View more View less

          Entrez Gene ID: (Human) 55512

          View more View less

          Function(s)
          phosphatidylserine binding catalytic activity sphingomyelin phosphodiesterase activity protein binding phosphoric diester hydrolase activity hydrolase activity identical protein binding metal ion binding neutral sphingomyelin phosphodiesterase activity phosphatidic acid binding
          Process(es)
          skeletal system development ossification endochondral ossification chondrocyte development hematopoietic progenitor cell differentiation regulation of leukocyte migration chondrocyte development involved in endochondral bone morphogenesis lipid metabolic process sphingolipid metabolic process ceramide metabolic process sphingomyelin metabolic process sphingomyelin catabolic process signal transduction polysaccharide transport peptide hormone secretion bone mineralization lung development BMP signaling pathway collagen metabolic process cellular response to reactive oxygen species multicellular organism growth positive regulation of mitotic nuclear division platelet-derived growth factor receptor signaling pathway lung alveolus development positive regulation of smooth muscle cell proliferation cartilage development negative regulation of protein kinase B signaling bone development respiratory system development regulation of cartilage development cellular response to hydrogen peroxide G1 to G0 transition cellular response to magnesium ion cellular response to tumor necrosis factor cellular response to redox state DNA biosynthetic process extracellular matrix assembly sphingolipid mediated signaling pathway dentinogenesis bone growth mitotic nuclear division cellular response to oxidised low-density lipoprotein particle stimulus regulation of hyaluronan biosynthetic process negative regulation of hyaluronan biosynthetic process cellular response to peptide positive regulation of exosomal secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline