Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MKAVKSERE RGSRRRHRDG DVVLPAGVVV KQERLSPEVA PPAHRRPDHS GGSPSPPTSE PARSGHRGNR ARGVSRSPPK KKNKASGRRS KSPRSKRNRS PHHSTVKVKQ EREDHPRRGR EDRQHREPSE QEHRRARNSD RDRHRGHSHQ RRTSNERPGS GQGQGRDRDT QNLQAQEEER EFYNARRREH RQRNDVGGGG SESQELVPRP GGNNKEKEVP AKEKPSFELS GALLEDTNTF RGVVIKYSEP PEARIPKKRW RLYPFKNDEV LPVMYIHRQS AYLLGRHRRI ADIPIDHPSC SKQHAVFQYR LVEYTRADGT VGRRVKPYII DLGSGNGTFL NNKRIEPQRY YELKEKDVLK FGFSSREYVL LHESSDTSEI DRKDDEDEEE EEEVSDS
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ423B22.2; FHA domain-containing protein SNIP1; PML1 homolog; RP3-423B22.3; Smad nuclear interacting protein 1 Snip1; SNIP1; unnamed protein product
Gene Aliases: 2410133M08Rik; NEDHCS; PML1; PMRED
UniProt ID: (Human) Q8TAD8, (Rat) Q5M9G6, (Mouse) Q8BIZ6
Entrez Gene ID: (Human) 79753, (Rat) 313588, (Mouse) 76793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support