Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MTVGKSSKM LQHIDYRMRC ILQDGRIFIG TFKAFDKHMN LILCDCDEFR KIKPKNAKQP EREEKRVLGL VLLRGENLVS MTVEGPPPKD TGIARVPLAG AAGGPGVGRA AGRGVPAGVP IPQAPAGLAG PVRGVGGPSQ QVMTPQGRGT VAAAAVAATA SIAGAPTQYP PGRGTPPPPV GRATPPPGIM APPPGMRPPM GPPIGLPPAR GTPIGMPPPG MRPPPPGIRG PPPPGMRPPR P (1-240 aa encoded by BC010057)
The protein encoded by this gene is one polypeptide of a small nuclear ribonucleoprotein complex and belongs to the snRNP SMB/SMN family. The protein plays a role in pre-mRNA processing, possibly tissue-specific alternative splicing events. Although individual snRNPs are believed to recognize specific nucleic acid sequences through RNA-RNA base pairing, the specific role of this family member is unknown. The protein arises from a bicistronic transcript that also encodes a protein identified as the SNRPN upstream reading frame (SNURF). Multiple transcription initiation sites have been identified and extensive alternative splicing occurs in the 5' untranslated region. Additional splice variants have been described but sequences for the complete transcripts have not been determined. The 5' UTR of this gene has been identified as an imprinting center. Alternative splicing or deletion caused by a translocation event in this paternally-expressed region is responsible for Angelman syndrome or Prader-Willi syndrome due to parental imprint switch failure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C0927; DKFZp686M12165; DKFZp761I1912; DKFZp762N022; FLJ33569; FLJ36996; FLJ39265; MGC29886; sm protein D; SM protein N; small nuclear ribonucleoprotein; small nuclear ribonucleoprotein particles component/spliceosome tissue-specific component; small ribonucleoprotein particle component; SmN; SNRPN; tissue-specific splicing protein; tissue-specific-splicing protein; unnamed protein product
Gene Aliases: 2410045I01Rik; HCERN3; Peg4; PWCR; RT-LI; SM-D; sm-N; SMN; SNRNP-N; SNURF-SNRPN
UniProt ID: (Human) P63162, (Mouse) P63163
Entrez Gene ID: (Human) 6638, (Mouse) 20646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support