Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SNX13 Antibodies

          Abnova

          SNX13 Polyclonal Antibody, MaxPab™

          View all (4) SNX13 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SNX13 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SNX13 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SNX13 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SNX13 Antibody (H00023161-B01P) in WB

          Western Blot analysis of SNX13 expression in transfected 293T cell line by SNX13 MaxPab polyclonal antibody. Lane 1: SNX13 transfected lysate(105.27 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SNX13 Antibody in Western Blot (WB)

          Product Details

          H00023161-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SNX13 (AAH45667.1, 1 a.a. approximately 957 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MLTEASLSIW GWGSLGIVLF LITFGPFVIF YLTFYILCFV GGGLVVTLLF GKTNSEKYLE QCEHSFLPPT SPGVPKCLEE MKREARTIKI DRRLTGANII DEPLQQVIQF SLRDYVQYWY YTLSDDESFL LEIRQTLQNA LIQFATRSKE IDWQPYFTTR IVDDFGTHLR VFRKAQQKIT EKDDQVKGTA EDLVDTFFEV EVEMEKEVCR DLVCTSPKDE EGFLRDLCEV LLYLLLPPGD FQNKIMRYFV REILARGILL PLINQLSDPD YINQYVIWMI RDSNCNYEAF MNIIKLSDNI GELEAVRDKA AEELQYLRSL DTAGDDINTI KNQINSLLFV KKVCDSRIQR LQSGKEINTV KLAANFGKLC TVPLDSILVD NVALQFFMDY MQQTGGQAHL FFWMTVEGYR VTAQQQLEVL LSRQRDGKHQ TNQTKGLLRA AAVGIYEQYL SEKASPRVTV DDYLVAKLAD TLNHEDPTPE IFDDIQRKVY ELMLRDERFY PSFRQNALYV RMLAELDMLK DPSFRGSDDG DGESFNGSPT GSINLSLDDL SNVSSDDSVQ LHAYISDTGV CNDHGKTYAL YAITVHRRNL NSEEMWKTYR RYSDFHDFHM RITEQFESLS SILKLPGKKT FNNMDRDFLE KRKKDLNAYL QLLLAPEMMK ASPALAHYVY DFLENKAYSK GKGDFARKMD TFVNPLRNSM RNVSNAVKSL PDNLAEGMTK MSDNMGKMSE RLGQDIKQSF FKVPPLIPKT DSDPEHRRVS AQLDDNVDDN IPLRVMLLLM DEVFDLKERN QWLRRNIKNL LQQLIRATYG DTINRKIVDH VDWMTSPEQV ADSVKRFRDA FWPNGILAEA VPCRDKSIRM RTRVAGKTKL LAIMPDELKH IIGAETTRKG ILRVFEMFQH NQLNRRMVYV FLEGFLETLF PQYKFRELFN KLHSRSKQMQ KYKQKLQTTQ APSLQKR

          Target Information

          May be involved in several stages of intracellular trafficking. May play a role in endosome homeostasis. Acts as a GAP for Galphas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: RGS domain- and PHOX domain-containing protein; RGS-PX1; Sorting nexin-13; unnamed protein product

          View more View less

          Gene Aliases: KIAA0713; RGS-PX1; SNX13

          View more View less

          UniProt ID: (Human) Q9Y5W8

          View more View less

          Entrez Gene ID: (Human) 23161

          View more View less

          Function(s)
          lipid binding phosphatidylinositol-3-phosphate binding phosphatidylinositol binding
          Process(es)
          intracellular protein transport negative regulation of signal transduction protein transport positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline