Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SOS1 Antibodies

          Invitrogen

          SOS1 Polyclonal Antibody

          View all (13) SOS1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SOS1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          SOS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SOS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SOS1 Antibody (PA5-105249) in WB

          Western blot analysis of SOS1 extracts from COLO205 cells (heat-shock treatment). Samples were incubated with SOS1 polyclonal antibody (Product # PA5-105249) using a dilution of 1:1,000. Observed bands: 150 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SOS1 Antibody in Western Blot (WB)
          SOS1 Antibody in Western Blot (WB)
          SOS1 Antibody in Western Blot (WB)
          SOS1 Antibody in Western Blot (WB)
          SOS1 Polyclonal Antibody

          Product Details

          PA5-105249

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human SOS1(Accession Q07889), corresponding to amino acid residues L1056-D1106.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2853855

          Product Specific Information

          Antibody detects endogenous levels of total SOS1.

          Target Information

          Son of sevenless (SOS) is a guanine nucleotide exchange factor that activates Ras in response to growth factor stimulation. Sos1, and its closely related Sos2 paralog, are comprised of an N-terminal histone-binding domain, a RacGEF catalytic (DH) domain juxtaposed with a PH domain, a Ras exchanger motif (REM) domain, a RasGEF catalytic domain, and a C-terminal proline-rich region. Sos1 catalyzes the GDP-GTP exchange within the membrane-bound GTPase Ras and thereby switches on a key signaling circuit that involves the activation of MAPK cascade central to cellular proliferation, survival and differentiation. Growth factor stimulation rapidly induces the phosphorylation of Sos on multiple serine and threonine sites. Phosphorylation of residues within the Sos C-terminus by the MAP/ERK kinase disrupts the Grb2-Sos complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G GINGF; gingival fibromatosis; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; guanine nucleotide releasing factor; ras guanine exchange factor; Son of sevenless homolog 1; son-of-sevenless 1; SOS-1; unnamed protein product

          View more View less

          Gene Aliases: 4430401P03Rik; 9630010N06; AI449023; GF1; GGF1; GINGF; HGF; NS4; SOS-1; SOS1

          View more View less

          UniProt ID: (Human) Q07889, (Mouse) Q62245

          View more View less

          Entrez Gene ID: (Human) 6654, (Rat) 313845, (Mouse) 20662

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity GTPase activator activity epidermal growth factor receptor binding protein binding SH3 domain binding protein kinase binding protein heterodimerization activity molecular condensate scaffold activity DNA binding Ras guanyl-nucleotide exchange factor activity Rho guanyl-nucleotide exchange factor activity guanyl-nucleotide exchange factor
          Process(es)
          B cell homeostasis hair follicle development lymphocyte homeostasis response to ischemia heart morphogenesis cardiac atrium morphogenesis pericardium morphogenesis regulation of transcription from RNA polymerase II promoter signal transduction epidermal growth factor receptor signaling pathway small GTPase mediated signal transduction Ras protein signal transduction vitellogenesis axon guidance insulin receptor signaling pathway fibroblast growth factor receptor signaling pathway Schwann cell development cytokine-mediated signaling pathway regulation of T cell differentiation in thymus multicellular organism growth Fc-epsilon receptor signaling pathway T cell activation regulation of cell proliferation regulation of T cell proliferation myelination positive regulation of epidermal growth factor receptor signaling pathway insulin-like growth factor receptor signaling pathway neurotrophin TRK receptor signaling pathway blood vessel morphogenesis B cell receptor signaling pathway leukocyte migration positive regulation of small GTPase mediated signal transduction palate development eyelid development in camera-type eye heart trabecula morphogenesis midbrain morphogenesis regulation of pro-B cell differentiation regulation of Rho protein signal transduction positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-78576c459-2wcz5:80/100.66.79.29:80.
          git-commit: 65c7ea78bfc366f1196f6925435a6e3b75c9e6fa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline