Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SPOCK1 Antibodies

          Osenses

          SPOCK Polyclonal Antibody

          1 Reference
          View all (6) SPOCK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SPOCK Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          OST00291W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:2,000
          -

          Immunohistochemistry (IHC)

          1:300-1:2,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from human SPOCK conjugated to blue carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat and shares 94% identity with mouse sequence.

          Specificity of this antibody: SPOCK1.

          Target Information

          SPOCK1, also known as Testican-1, is a gene located on chromosome 5q31.2, encoding a member of the proteoglycan family characterized by containing multiple domains that facilitate interactions with other extracellular matrix proteins. SPOCK1 is primarily expressed in various tissues, including the brain and reproductive organs, where it plays crucial roles in modulating cell adhesion, migration, and matrix remodeling. Functionally, SPOCK1 contributes to the regulation of extracellular matrix composition, influencing processes such as tissue development and wound healing. In the context of cancer, SPOCK1 is often upregulated, promoting tumor progression and metastasis by enhancing cancer cell motility and invasiveness. Its involvement in these pathological mechanisms makes SPOCK1 a potential biomarker for cancer prognosis and a target for therapeutic intervention. Research continues to unravel SPOCK1's diverse biological activities and its impact on cellular architecture and disease progression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ37170; Protein SPOCK; SPARC/osteonectin, cwcv and kazal like domains proteoglycan 1; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1; Testican-1; unnamed protein product

          View more View less

          Gene Aliases: SPOCK; SPOCK1; TESTICAN; TIC1; TICN1

          View more View less

          UniProt ID: (Human) Q08629

          View more View less

          Entrez Gene ID: (Human) 6695

          View more View less

          Function(s)
          serine-type endopeptidase inhibitor activity cysteine-type endopeptidase inhibitor activity extracellular matrix structural constituent calcium ion binding protein binding collagen binding metalloendopeptidase inhibitor activity extracellular matrix binding
          Process(es)
          regulation of cell growth neuron migration cell adhesion nervous system development negative regulation of cell-substrate adhesion negative regulation of neuron projection development central nervous system neuron differentiation neurogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0