Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SPOCK1 Antibodies

          Abnova

          SPOCK1 Polyclonal Antibody, MaxPab™

          View all (6) SPOCK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SPOCK1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SPOCK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SPOCK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SPOCK1 Antibody (H00006695-B02P) in WB

          Western Blot analysis of SPOCK1 expression in transfected 293T cell line by SPOCK1 MaxPab polyclonal antibody. Lane 1: SPOCK1 transfected lysate(48.29 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SPOCK1 Antibody in Western Blot (WB)

          Product Details

          H00006695-B02P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SPOCK1 (AAH30691.1, 1 a.a. approximately 439 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPAIAVLAAA AAAWCFLQVE SRHLDALAGG AGPNHGNFLD NDQWLSTVSQ YDRDKYWNRF RDDDYFRNWN PNKPFDKALD PSKDPCLKVK CSPHKVCVTQ DYQTALCVSR KHLLPRQKKG NVAQKHWVGP SNLVKCKPCP VAQSAMVCGS DGHSYTSKCK LEFHACSTGK SLATLCDGPC PCLPEPEPPK HKAERSACTD KELRNLASRL KDWFGALHED ANRVIKPTSS NTAQGRFDTS ILPICKDSLG WMFNKLDMNY DLLLDPSEIN AIYLDKYEPC IKPLFNSCDS FKDGKLSNNE WCYCFQKPGG LPCQNEMNRI QKLSKGKSLL GAFIPRCNEE GYYKATQCHG STGQCWCVDK YGNELAGSRK QGAVSCEEEQ ETSGDFGSGG SVVLLDDLEY ERELGPKDKE GKLRVHTRAV TEDDEDEDDD KEDEVGYIW

          Target Information

          SPOCK1, also known as Testican-1, is a gene located on chromosome 5q31.2, encoding a member of the proteoglycan family characterized by containing multiple domains that facilitate interactions with other extracellular matrix proteins. SPOCK1 is primarily expressed in various tissues, including the brain and reproductive organs, where it plays crucial roles in modulating cell adhesion, migration, and matrix remodeling. Functionally, SPOCK1 contributes to the regulation of extracellular matrix composition, influencing processes such as tissue development and wound healing. In the context of cancer, SPOCK1 is often upregulated, promoting tumor progression and metastasis by enhancing cancer cell motility and invasiveness. Its involvement in these pathological mechanisms makes SPOCK1 a potential biomarker for cancer prognosis and a target for therapeutic intervention. Research continues to unravel SPOCK1's diverse biological activities and its impact on cellular architecture and disease progression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ37170; Protein SPOCK; SPARC/osteonectin, cwcv and kazal like domains proteoglycan 1; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1; Testican-1; unnamed protein product

          View more View less

          Gene Aliases: SPOCK; SPOCK1; TESTICAN; TIC1; TICN1

          View more View less

          UniProt ID: (Human) Q08629

          View more View less

          Entrez Gene ID: (Human) 6695

          View more View less

          Function(s)
          serine-type endopeptidase inhibitor activity cysteine-type endopeptidase inhibitor activity extracellular matrix structural constituent calcium ion binding protein binding collagen binding metalloendopeptidase inhibitor activity extracellular matrix binding
          Process(es)
          regulation of cell growth neuron migration cell adhesion nervous system development negative regulation of cell-substrate adhesion negative regulation of neuron projection development central nervous system neuron differentiation neurogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline