Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SRC Antibodies

          Invitrogen

          SRC Polyclonal Antibody

          Advanced Verification
          5 Published Figures
          14 References
          View all (73) SRC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SRC Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (5)
          • Advanced Verification (1)
          SRC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SRC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRC Antibody (44-656G) in WB

          Western Blot. Analysis Extracts of CEFs untransfected (-) or transfected (+) with wild-type human Src were resolved by SDS-PAGE on a 10% Tris-glycine gel and transferred to nitrocellulose. The membrane was blocked with a 5% BSA-TBST buffer overnight at 4°C, then incubated with 0.5 µg/mL Src pan antibody for two hours at room temperature in a 3% BSA-TBST buffer. After washing, the membrane was incubated with goat F (ab')2 anti-rabbit IgG alkaline phosphatase (Product # ALI4405) and signals were detected using the Tropix WesternStar™ method.... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRC Antibody in Western Blot (WB)
          SRC Antibody in Western Blot (WB)
          SRC Antibody in Western Blot (WB)
          SRC Antibody in Western Blot (WB)
          SRC Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SRC Antibody in Immunoprecipitation (IP)
          SRC Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SRC Polyclonal Antibody

          Product Details

          44-656G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 10 publications 10 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized peptide derived from the amino acid region 31-49 of human Src protein. The sequence is conserved in mouse and rat.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533712

          Target Information

          SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth. SRC is involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: EC 2.7.10.2; neuronal proto-oncogene tyrosine-protein kinase Src; OTTHUMP00000174476; OTTHUMP00000174477; OTTHUMP00000174478; p60-Src; pp60src; proto-oncogene c-Src; protooncogene SRC, Rous sarcoma; Rous sarcoma oncogene; RP5-823N20.1; tyrosine kinase pp60c-src; tyrosine-protein kinase SRC-1; tyrosine-specific protein kinase; unnamed protein product; v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog

          View more View less

          Gene Aliases: ASV; c-SRC; p60-Src; pp60c-src; SRC1; THC6

          View more View less

          UniProt ID: (Human) P12931, (Mouse) P05480

          View more View less

          Entrez Gene ID: (Human) 6714, (Mouse) 20779

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding integrin binding protein binding ATP binding phospholipase activator activity kinase activity enzyme binding protein domain specific binding heme binding protein tyrosine kinase activator activity receptor activator activity ionotropic glutamate receptor binding SH2 domain binding phospholipase binding ion channel binding cadherin binding ephrin receptor binding ATPase binding phosphoprotein binding BMP receptor binding connexin binding scaffold protein binding protein kinase C binding insulin receptor binding protein kinase binding estrogen receptor binding macromolecular complex binding cell adhesion molecule binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter primary ovarian follicle growth positive regulation of cytokine production protein phosphorylation cell adhesion epidermal growth factor receptor signaling pathway transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway gamete generation lactation positive regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression regulation of epithelial cell migration positive regulation of epithelial cell migration positive regulation of glucose metabolic process positive regulation of protein processing positive regulation of smooth muscle cell migration cell migration protein deubiquitination peptidyl-tyrosine phosphorylation regulation of cell-cell adhesion cell differentiation positive regulation of cell migration forebrain development negative regulation of protein complex assembly protein destabilization cellular response to nutrient levels negative regulation of telomere maintenance regulation of intracellular estrogen receptor signaling pathway toll-like receptor 3 signaling pathway regulation of toll-like receptor 3 signaling pathway substrate adhesion-dependent cell spreading cellular response to reactive oxygen species positive regulation of Rac protein signal transduction positive regulation of dephosphorylation negative regulation of hippo signaling intracellular signal transduction osteoclast development cellular response to platelet-derived growth factor stimulus peptidyl-tyrosine autophosphorylation angiotensin-activated signaling pathway TORC1 signaling vasodilation odontogenesis positive regulation of apoptotic process negative regulation of apoptotic process stress fiber assembly proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of MAP kinase activity transcytosis bone resorption positive regulation of Notch signaling pathway positive regulation of bone resorption positive regulation of cell adhesion positive regulation of cell size positive regulation of glycolytic process positive regulation of Ras protein signal transduction positive regulation of insulin receptor signaling pathway neurotrophin TRK receptor signaling pathway focal adhesion assembly oogenesis positive regulation of peptidyl-tyrosine phosphorylation progesterone receptor signaling pathway positive regulation of small GTPase mediated signal transduction positive regulation of protein transport negative regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of mitochondrial depolarization uterus development branching involved in mammary gland duct morphogenesis regulation of cell projection assembly intestinal epithelial cell development interleukin-6-mediated signaling pathway cellular response to hydrogen peroxide positive regulation of ERK1 and ERK2 cascade response to interleukin-1 cellular response to peptide hormone stimulus cellular response to progesterone stimulus cellular response to fluid shear stress cellular response to transforming growth factor beta stimulus positive regulation of podosome assembly DNA biosynthetic process regulation of heart rate by cardiac conduction positive regulation of canonical Wnt signaling pathway positive regulation of wound healing positive regulation of protein localization to nucleus positive regulation of intracellular signal transduction negative regulation of neutrophil activation positive regulation of cellular protein localization negative regulation of TORC1 signaling positive regulation of TORC1 signaling cellular response to angiotensin positive regulation of vascular smooth muscle cell proliferation positive regulation of neural precursor cell proliferation positive regulation of male germ cell proliferation positive regulation of ovarian follicle development positive regulation of lamellipodium morphogenesis positive regulation of platelet-derived growth factor receptor-beta signaling pathway regulation of early endosome to late endosome transport negative regulation of anoikis negative regulation of extrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway regulation of caveolin-mediated endocytosis stimulatory C-type lectin receptor signaling pathway negative regulation of inflammatory response to antigenic stimulus signal transduction signal complex assembly macroautophagy platelet activation osteoclast differentiation T cell costimulation positive regulation of integrin activation Fc-gamma receptor signaling pathway involved in phagocytosis ERBB2 signaling pathway regulation of vascular permeability regulation of bone resorption viral entry into host cell vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway leukocyte migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0