Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SSH1 Antibodies

          Abnova

          SSH1 Polyclonal Antibody, MaxPab™

          View all (15) SSH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SSH1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SSH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SSH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SSH1 Antibody (H00054434-B01P) in WB

          Western Blot analysis of SSH1 expression in transfected 293T cell line by SSH1 MaxPab polyclonal antibody. Lane1:SSH1 transfected lysate(115.39 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SSH1 Antibody in Western Blot (WB)

          Product Details

          H00054434-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SSH1 (NP_061857.2, 1 a.a. approximately 1049 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVLVTLQRSP TPSAASSSAS NSELEAGSEE DRKLNLSLSE SFFMVKGAAL FLQQGSSPQG QRSLQHPHKH AGDLPQHLQV MINLLRCEDR IKLAVRLESA WADRVRYMVV VYSSGRQDTE ENILLGVDFS SKESKSCTIG MVLRLWSDTK IHLDGDGGFS VSTAGRMHIF KPVSVQAMWS ALQVLHKACE VARRHNYFPG GVALIWATYY ESCISSEQSC INEWNAMQDL ESTRPDSPAL FVDKPTEGER TERLIKAKLR SIMMSQDLEN VTSKEIRNEL EKQMNCNLKE LKEFIDNEML LILGQMDKPS LIFDHLYLGS EWNASNLEEL QGSGVDYILN VTREIDNFFP GLFAYHNIRV YDEETTDLLA HWNEAYHFIN KAKRNHSKCL VHCKMGVSRS ASTVIAYAMK EFGWPLEKAY NYVKQKRSIT RPNAGFMRQL SEYEGILDAS KQRHNKLWRQ QTDSSLQQPV DDPAGPGDFL PETPDGTPES QLPFLDDAAQ PGLGPPLPCC FRRLSDPLLP SPEDETGSLV HLEDPEREAL LEEAAPPAEV HRPARQPQQG SGLCEKDVKK KLEFGSPKGR SGSLLQVEET EREEGLGAGR WGQLPTQLDQ NLLNSENLNN NSKRSCPNGM EDDAIFGILN KVKPSYKSCA DCMYPTASGA PEASRERCED PNAPAICTQP AFLPHITSSP VAHLASRSRV PEKPASGPTE PPPFLPPAGS RRADTSGPGA GAALEPPASL LEPSRETPKV LPKSLLLKNS HCDKNPPSTE VVIKEESSPK KDMKPAKDLR LLFSNESEKP TTNSYLMQHQ ESIIQLQKAG LVRKHTKELE RLKSVPADPA PPSRDGPASR LEASIPEESQ DPAALHELGP LVMPSQAGSD EKSEAAPASL EGGSLKSPPP FFYRLDHTSS FSKDFLKTIC YTPTSSSMSS NLTRSSSSDS IHSVRGKPGL VKQRTQEIET RLRLAGLTVS SPLKRSHSLA KLGSLTFSTE DLSSEADPST VADSQDTTLS ESSFLHEPQG TPRDPAATSK PSGKPAPENL KSPSWMSKS

          Target Information

          This gene encodes a member of the SWI/SNF family, whose members have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. It possesses at least two conserved domains that could be important for its function. First, it has a DNA-binding domain that can specifically bind an AT-rich DNA sequence known to be recognized by a SNF/SWI complex at the beta-globin locus. Second, the C-terminus of the protein can stimulate glucocorticoid receptor-dependent transcriptional activation. It is thought that the protein encoded by this gene confers specificity to the SNF/SWI complex and may recruit the complex to its targets through either protein-DNA or protein-protein interactions. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hSSH-1L; Protein phosphatase Slingshot homolog 1; slingshot homolog 1; SSH-1L; SSH-like protein 1; unnamed protein product

          View more View less

          Gene Aliases: KIAA1298; SSH1; SSH1L

          View more View less

          UniProt ID: (Human) Q8WYL5

          View more View less

          Entrez Gene ID: (Human) 54434

          View more View less

          Function(s)
          actin binding phosphoprotein phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding hydrolase activity phosphatase activity
          Process(es)
          cell morphogenesis protein dephosphorylation actin cytoskeleton organization negative regulation of actin filament polymerization positive regulation of synaptic plasticity regulation of protein metabolic process cellular response to ATP excitatory chemical synaptic transmission positive regulation of AMPA glutamate receptor clustering positive regulation of vascular associated smooth muscle cell migration positive regulation of excitatory postsynaptic potential
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline