Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • ST3GAL3 Antibodies

          Invitrogen

          ST3GAL3 Polyclonal Antibody

          View all (6) ST3GAL3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ST3GAL3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          ST3GAL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ST3GAL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ST3GAL3 Antibody (PA5-110436) in ICC/IF

          Immunofluorescence analysis of ST3GAL3 in HeLa cells. Samples were incubated with ST3GAL3 Polyclonal antibody (Product # PA5-110436). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ST3GAL3 Antibody in Immunocytochemistry (ICC/IF)
          ST3GAL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST3GAL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST3GAL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST3GAL3 Antibody in Western Blot (WB)
          ST3GAL3 Polyclonal Antibody

          Product Details

          PA5-110436

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 30-375 of human ST3GAL3 (NP_006270.1).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.52 mg/mL

          Amount

          52 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2855847

          Product Specific Information

          Immunogen sequence: LHLLQWEEDS NSVVLSFDSA GQTLGSEYDR LGFLLNLDSK LPAELATKYA NFSEGACKPG YASALMTAIF PRFSKPAPMF LDDSFRKWAR IREFVPPFGI KGQDNLIKAI LSVTKEYRLT PALDSLRCRR CIIVGNGGVL ANKSLGSRID DYDIVVRLNS APVKGFEKDV GSKTTLRITY PEGAMQRPEQ YERDSLFVLA GFKWQDFKWL KYIVYKERVS ASDGFWKSVA TRVPKEPPEI RILNPYFIQE AAFTLIGLPF NNGLMGRGNI PTLGSVAVTM ALHGCDEVAV AGFGYDMSTP NAPLHYYETV RMAAIKESWT HNIQREKEFL RKLVKARVIT DLSSGI

          Target Information

          The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha 2,3-sialyltransferase III; Alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; Beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; Sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6 (N-acetyllactosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); ST3Gal III; ST3GalIII; ST3N; unnamed protein product

          View more View less

          Gene Aliases: DEE15; EIEE15; MRT12; Siat3; SIAT6; ST3Gal III; ST3GAL3; ST3GALII; ST3GalIII; ST3N

          View more View less

          UniProt ID: (Human) Q11203, (Mouse) P97325, (Rat) Q02734

          View more View less

          Entrez Gene ID: (Human) 6487, (Mouse) 20441, (Rat) 64445

          View more View less

          Function(s)
          beta-galactoside (CMP) alpha-2,3-sialyltransferase activity protein binding N-acetyllactosaminide alpha-2,3-sialyltransferase activity sialyltransferase activity transferase activity transferase activity, transferring glycosyl groups transferase metabolite interconversion enzyme
          Process(es)
          protein glycosylation glycosphingolipid biosynthetic process oligosaccharide biosynthetic process ganglioside biosynthetic process via lactosylceramide O-glycan processing keratan sulfate biosynthetic process viral protein processing sialylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0