Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • STK36 Antibodies

          Abnova

          STK36 Polyclonal Antibody, MaxPab™

          View all (9) STK36 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite STK36 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          STK36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          STK36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          STK36 Antibody (H00027148-B01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to STK36 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          STK36 Antibody in Immunocytochemistry (ICC/IF)
          STK36 Antibody in Western Blot (WB)
          STK36 Polyclonal Antibody, MaxPab™

          Product Details

          H00027148-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          STK36 (NP_056505.1, 1 a.a. approximately 1315 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MEKYHVLEMI GEGSFGRVYK GRRKYSAQVV ALKFIPKLGR SEKELRNLQR EIEIMRGLRH PNIVHMLDSF ETDKEVVVVT DYAEGELFQI LEDDGKLPED QVQAIAAQLV SALYYLHSHR ILHRDMKPQN ILLAKGGGIK LCDFGFARAM STNTMVLTSI KGTPLYMSPE LVEERPYDHT ADLWSVGCIL YELAVGTPPF YATSIFQLVS LILKDPVRWP STISPCFKDF LQGLLTKDPR QRLSWPDLLY HPFIAGHVTI ITEPAGPDLG TPFTSRLPPE LQVLKDEQAH RLAPKGNQSR ILTQAYKRMA EEAMQKKHQN TGPALEQEDK TSKVAPGTAP LPRLGATPQE SSLLAGILAS ELKSSWAKSG TGEVPSAPRE NRTTPDCERA FPEERPEVLG QRSTDVVDLE NEEPDSDNEW QHLLETTEPV PIQLKAPLTL LCNPDFCQRI QSQLHEAGGQ ILNGILEGAS HILPAFRVLS SLLSSCSDSV ALYSFCREAG LPGLLLSLLR HSQESNSLQQ QSWYGTFLQD LMAVIQAYFA CTFNLERSQT SDSLQVFQEA ANLFLDLLGK LLAQPDDSEQ TLRRDSLMCF TVLCEAMDGN SRAISKAFYS SLLTTQQVVL DGLLHGLTVP QLPVHTPQGA PQVSQPLREQ SEDIPGAISS ALAAICTAPV GLPDCWDAKE QVCWHLANQL TEDSSQLRPS LISGLQHPIL CLHLLKVLYS CCLVSEGLCR LLGQEPLALE SLFMLIQGKV KVVDWEESTE VTLYFLTLLV FRLQNLPCGM EKLGSDVATL FTHSHVVSLV SAAACLLGQL GQQGVTFDLQ PMEWMAAATH ALSAPAEVRL TPPGSCGFYD GLLILLLQLL TEQGKASLIR DMSSSEMWTV LWHRFSMVLR LPEEASAQEG ELSLSSPPSP EPDWTLISPQ GMAALLSLAM ATFTQEPQLC LSCLSQHGSI LMSILKHLLC PSFLNQLRQA PHGSEFLPVV VLSVCQLLCF PFALDMDADL LIGVLADLRD SEVAAHLLQV CCYHLPLMQV ELPISLLTRL ALMDPTSLNQ FVNTVSASPR TIVSFLSVAL LSDQPLLTSD LLSLLAHTAR VLSPSHLSFI QELLAGSDES YRPLRSLLGH PENSVRAHTY RLLGHLLQHS MALRGALQSQ SGLLSLLLLG LGDKDPVVRC SASFAVGNAA YQAGPLGPAL AAAVPSMTQL LGDPQAGIRR NVASALGNLG PEGLGEELLQ CEVPQRLLEM ACGDPQPNVK EAALIALRSL QQEPGIHQVL VSLGASEKLS LLSLGNQSLP HSSPRPASAK HCRKLIHLLR PAHSM

          Target Information

          Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Fused homolog; Serine/threonine-protein kinase 36; testicular tissue protein Li 188; unnamed protein product

          View more View less

          Gene Aliases: CILD46; FU; KIAA1278; STK36

          View more View less

          UniProt ID: (Human) Q9NRP7

          View more View less

          Entrez Gene ID: (Human) 27148

          View more View less

          Function(s)
          nucleotide binding transcription corepressor binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity metal ion binding protein serine kinase activity
          Process(es)
          epithelial cilium movement protein phosphorylation microtubule-based process smoothened signaling pathway brain development post-embryonic development cell projection organization axoneme assembly positive regulation of smoothened signaling pathway regulation of sequence-specific DNA binding transcription factor activity cilium morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline