Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • STK36 Antibodies

          Abnova

          STK36 Polyclonal Antibody, MaxPab™

          View all (8) STK36 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite STK36 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          STK36 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          STK36 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          STK36 Antibody (H00027148-D01P) in WB

          STK36 MaxPab rabbit polyclonal antibody. Western Blot analysis of STK36 expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          STK36 Antibody in Western Blot (WB)

          Product Details

          H00027148-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:50-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          STK36 (NP_056505.1, 1 a.a. approximately 1315 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEKYHVLEMI GEGSFGRVYK GRRKYSAQVV ALKFIPKLGR SEKELRNLQR EIEIMRGLRH PNIVHMLDSF ETDKEVVVVT DYAEGELFQI LEDDGKLPED QVQAIAAQLV SALYYLHSHR ILHRDMKPQN ILLAKGGGIK LCDFGFARAM STNTMVLTSI KGTPLYMSPE LVEERPYDHT ADLWSVGCIL YELAVGTPPF YATSIFQLVS LILKDPVRWP STISPCFKDF LQGLLTKDPR QRLSWPDLLY HPFIAGHVTI ITEPAGPDLG TPFTSRLPPE LQVLKDEQAH RLAPKGNQSR ILTQAYKRMA EEAMQKKHQN TGPALEQEDK TSKVAPGTAP LPRLGATPQE SSLLAGILAS ELKSSWAKSG TGEVPSAPRE NRTTPDCERA FPEERPEVLG QRSTDVVDLE NEEPDSDNEW QHLLETTEPV PIQLKAPLTL LCNPDFCQRI QSQLHEAGGQ ILNGILEGAS HILPAFRVLS SLLSSCSDSV ALYSFCREAG LPGLLLSLLR HSQESNSLQQ QSWYGTFLQD LMAVIQAYFA CTFNLERSQT SDSLQVFQEA ANLFLDLLGK LLAQPDDSEQ TLRRDSLMCF TVLCEAMDGN SRAISKAFYS SLLTTQQVVL DGLLHGLTVP QLPVHTPQGA PQVSQPLREQ SEDIPGAISS ALAAICTAPV GLPDCWDAKE QVCWHLANQL TEDSSQLRPS LISGLQHPIL CLHLLKVLYS CCLVSEGLCR LLGQEPLALE SLFMLIQGKV KVVDWEESTE VTLYFLTLLV FRLQNLPCGM EKLGSDVATL FTHSHVVSLV SAAACLLGQL GQQGVTFDLQ PMEWMAAATH ALSAPAEVRL TPPGSCGFYD GLLILLLQLL TEQGKASLIR DMSSSEMWTV LWHRFSMVLR LPEEASAQEG ELSLSSPPSP EPDWTLISPQ GMAALLSLAM ATFTQEPQLC LSCLSQHGSI LMSILKHLLC PSFLNQLRQA PHGSEFLPVV VLSVCQLLCF PFALDMDADL LIGVLADLRD SEVAAHLLQV CCYHLPLMQV ELPISLLTRL ALMDPTSLNQ FVNTVSASPR TIVSFLSVAL LSDQPLLTSD LLSLLAHTAR VLSPSHLSFI QELLAGSDES YRPLRSLLGH PENSVRAHTY RLLGHLLQHS MALRGALQSQ SGLLSLLLLG LGDKDPVVRC SASFAVGNAA YQAGPLGPAL AAAVPSMTQL LGDPQAGIRR NVASALGNLG PEGLGEELLQ CEVPQRLLEM ACGDPQPNVK EAALIALRSL QQEPGIHQVL VSLGASEKLS LLSLGNQSLP HSSPRPASAK HCRKLIHLLR PAHSM

          Target Information

          Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Fused homolog; serine/threonine kinase 36 (fused homolog, Drosophila); Serine/threonine-protein kinase 36; stk36; testicular tissue protein Li 188; unnamed protein product

          View more View less

          Gene Aliases: 1700112N14Rik; B930045J24; CILD46; FU; Fused; KIAA1278; mKIAA1278; STK36

          View more View less

          UniProt ID: (Human) Q9NRP7, (Mouse) Q69ZM6

          View more View less

          Entrez Gene ID: (Human) 27148, (Mouse) 269209

          View more View less

          Function(s)
          nucleotide binding transcription corepressor binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity metal ion binding protein serine kinase activity transcription factor binding transferase activity, transferring phosphorus-containing groups non-receptor serine/threonine protein kinase protein modifying enzyme
          Process(es)
          epithelial cilium movement protein phosphorylation microtubule-based process smoothened signaling pathway brain development post-embryonic development cell projection organization axoneme assembly positive regulation of smoothened signaling pathway regulation of sequence-specific DNA binding transcription factor activity cilium morphogenesis positive regulation of hh target transcription factor activity multicellular organism development phosphorylation cilium assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline