Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SULF1 Antibodies

          Abnova

          SULF1 Polyclonal Antibody, MaxPab™

          View all (8) SULF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SULF1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SULF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SULF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SULF1 Antibody (H00023213-D01P) in WB

          Western Blot analysis of SULF1 expression in transfected 293T cell line by SULF1 MaxPab polyclonal antibody. Lane 1: SULF1 transfected lysate(101.00 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SULF1 Antibody in Western Blot (WB)
          SULF1 Polyclonal Antibody, MaxPab™

          Product Details

          H00023213-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SULF1 (NP_055985.1, 1 a.a. approximately 871 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MKYSCCALVL AVLGTELLGS LCSTVRSPRF RGRIQQERKN IRPNIILVLT DDQDVELGSL QVMNKTRKIM EHGGATFINA FVTTPMCCPS RSSMLTGKYV HNHNVYTNNE NCSSPSWQAM HEPRTFAVYL NNTGYRTAFF GKYLNEYNGS YIPPGWREWL GLIKNSRFYN YTVCRNGIKE KHGFDYAKDY FTDLITNESI NYFKMSKRMY PHRPVMMVIS HAAPHGPEDS APQFSKLYPN ASQHITPSYN YAPNMDKHWI MQYTGPMLPI HMEFTNILQR KRLQTLMSVD DSVERLYNML VETGELENTY IIYTADHGYH IGQFGLVKGK SMPYDFDIRV PFFIRGPSVE PGSIVPQIVL NIDLAPTILD IAGLDTPPDV DGKSVLKLLD PEKPGNRFRT NKKAKIWRDT FLVERGKFLR KKEESSKNIQ QSNHLPKYER VKELCQQARY QTACEQPGQK WQCIEDTSGK LRIHKCKGPS DLLTVRQSTR NLYARGFHDK DKECSCRESG YRASRSQRKS QRQFLRNQGT PKYKPRFVHT RQTRSLSVEF EGEIYDINLE EEEELQVLQP RNIAKRHDEG HKGPRDLQAS SGGNRGRMLA DSSNAVGPPT TVRVTHKCFI LPNDSIHCER ELYQSARAWK DHKAYIDKEI EALQDKIKNL REVRGHLKRR KPEECSCSKQ SYYNKEKGVK KQEKLKSHLH PFKEAAQEVD SKLQLFKENN RRRKKERKEK RRQRKGEECS LPGLTCFTHD NNHWQTAPFW NLGSFCACTS SNNNTYWCLR TVNETHNFLF CEFATGFLEY FDMNTDPYQL TNTVHTVERG ILNQLHVQLM ELRSCQGYKQ CNPRPKNLDV GNKDGGSYDL HRGQLWDGWE G

          Target Information

          Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Arylsulfatase; Extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; N-acetylglucosamine-6-sulfatase; sulfatase FP; unnamed protein product

          View more View less

          Gene Aliases: KIAA1077; SULF-1; SULF1

          View more View less

          UniProt ID: (Human) Q8IWU6

          View more View less

          Entrez Gene ID: (Human) 23213

          View more View less

          Function(s)
          arylsulfatase activity calcium ion binding glycosaminoglycan binding N-acetylglucosamine-6-sulfatase activity sulfuric ester hydrolase activity hydrolase activity metal ion binding
          Process(es)
          kidney development negative regulation of endothelial cell proliferation chondrocyte development glomerular filtration apoptotic process positive regulation of vascular endothelial growth factor production esophagus smooth muscle contraction negative regulation of angiogenesis positive regulation of Wnt signaling pathway heparan sulfate proteoglycan metabolic process negative regulation of cell migration positive regulation of BMP signaling pathway glomerular basement membrane development glial cell-derived neurotrophic factor receptor signaling pathway regulation of fibroblast growth factor receptor signaling pathway negative regulation of fibroblast growth factor receptor signaling pathway vascular endothelial growth factor receptor signaling pathway embryonic skeletal system development cartilage development bone development innervation negative regulation of prostatic bud formation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline