Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SULT1 Family Antibodies

          Bioss

          SULT1 Family Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SULT1 Family Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          BS-6283R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human SULT1A1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase 1A3/1A4; Aryl sulfotransferase 2; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; Aryl sulfotransferase IV; arylamine sulfotransferase; arylsulfotransferase ST1A4; ASTIV; Catecholamine-sulfating phenol sulfotransferase; cytosolic sulfotransferase; DOPA/tyrosine sulfotransferase; dopamine-specific sulfotransferase; HAST-I; HAST1/HAST2; HAST3; M-PST; Minoxidil sulfotransferase; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; mSTp1; Mx-ST; N-hydroxyarylamine sulfotransferase; N-hydroxylarylamine sulfotransferase; P-PST 1; P-PST 2; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol sulfotransferase 1A5; Phenol sulfotransferase 1c1; Phenol sulfotransferase 2; phenol-preferring phenol sulfotransferase2; Phenol-sulfating phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 2; phenol/aryl sulfotransferase; phenolic-metabolizing (P) form of PST; Placental estrogen sulfotransferase; PST-1; SLX1B-SULT1A4; ST1A1; ST1A2; ST1A3; ST1A4; ST1B1; ST1C1; ST1C3; ST1C4; Sulfokinase; Sulfotransferase 1A1; Sulfotransferase 1A2; Sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; Sulfotransferase 1B1; Sulfotransferase 1B2; Sulfotransferase 1C1; Sulfotransferase 1C2; Sulfotransferase 1C3; Sulfotransferase 1C4; sulfotransferase family 1A phenol-preferring member 2; sulfotransferase family 1A, member 2; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family 1A, phenol-preferring, member 2; sulfotransferase family 1B, member 1; Sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4; sulfotransferase family, cytosolic, 1B, member 1; sulfotransferase family, cytosolic, 1C, member 1; sulfotransferase family, cytosolic, 1C, member 2; sulfotransferase family, cytosolic, 1C, member 3; sulfotransferase family, cytosolic, 1C, member 4; sulfotransferase family, cytosolic, 1C, member C2; Sulfotransferase, monoamine-preferring; sulfotransferase, phenol preferring 1; sulfotransferase, phenol preferring 2; sulphotransferase family cytosolic 1A phenol-prefering member 1; sulphotransferase family cytosolic 1A phenol-preferring member 2; SULT1C#2; sult1c1; thermolabile (monoamine, M form) phenol sulfotransferase; Thermolabile phenol sulfotransferase; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; Thyroid hormone sulfotransferase; TL-PST; Ts-PST; Tyrosine-ester sulfotransferase; unnamed protein product

          View more View less

          Gene Aliases: (PST)G; AI266890; ASTIV; HAST; HAST-I; HAST1/HAST2; HAST3; HAST4; M-PST; mOLFST; Mx-ST; OK/SW-cl.88; P-PST; P-PST 1; P-PST 2; P-SULT; PST; PST-1; ST1A1; ST1A2; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; ST1B1; ST1B2; ST1C1; ST1C3; St2b2; STM; STP; STP1; STP2; SULT1A1; SULT1A2; SULT1A3; SULT1A4; SULT1B1; SULT1B2; SULT1C; Sult1c1; SULT1C2; SULT1C3; SULT1C4; TL-PST; ts-PST; TSPST1; TSPST2

          View more View less

          UniProt ID: (Human) P50225, (Human) P50226, (Human) P0DMM9, (Human) O43704, (Human) Q6IMI6, (Human) O75897, (Rat) P17988, (Mouse) Q9QWG7, (Rat) P52847, (Mouse) Q80VR3, (Rat) P50237

          View more View less

          Entrez Gene ID: (Human) 6817, (Human) 6799, (Human) 6818, (Human) 445329, (Human) 27284, (Human) 442038, (Human) 27233, (Rat) 83783, (Mouse) 20887, (Mouse) 56362, (Rat) 64305, (Mouse) 20888, (Rat) 65185

          View more View less

          Function(s)
          aryl sulfotransferase activity protein binding sulfotransferase activity transferase activity flavonol 3-sulfotransferase activity steroid sulfotransferase activity 3'-phosphoadenosine 5'-phosphosulfate binding sulfate binding amine sulfotransferase activity alcohol sulfotransferase activity bile-salt sulfotransferase activity nucleotide binding protein homodimerization activity N-acetylglucosamine 6-O-sulfotransferase activity N-acetylgalactosamine 4-O-sulfotransferase activity heparan sulfate 2-O-sulfotransferase activity HNK-1 sulfotransferase activity tyrosine-ester sulfotransferase activity heparan sulfate 6-O-sulfotransferase activity trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity 1-phenanthrol sulfotransferase activity 3-phenanthrol sulfotransferase activity 4-phenanthrol sulfotransferase activity trans-3,4-dihydrodiolphenanthrene sulfotransferase activity 9-phenanthrol sulfotransferase activity 2-phenanthrol sulfotransferase activity phenanthrol sulfotransferase activity 1-hydroxypyrene sulfotransferase activity galactose 3-O-sulfotransferase activity proteoglycan sulfotransferase activity cholesterol sulfotransferase activity hydroxyjasmonate sulfotransferase activity transferase
          Process(es)
          ethanol catabolic process catecholamine metabolic process lipid metabolic process xenobiotic metabolic process steroid metabolic process estrogen metabolic process amine metabolic process flavonoid metabolic process thyroid hormone metabolic process dopamine catabolic process 3'-phosphoadenosine 5'-phosphosulfate metabolic process sulfation amine biosynthetic process phenol-containing compound metabolic process epinephrine metabolic process norepinephrine metabolic process dopamine metabolic process serotonin metabolic process cellular response to dopamine cellular biogenic amine metabolic process epithelial cell differentiation sulfur compound metabolic process cholesterol metabolic process doxorubicin metabolic process regulation of blood pressure response to activity response to insecticide drug metabolic process 4-nitrophenol metabolic process response to glucocorticoid metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline