FIGURE: 1 / 2
PA1-31763 detects GCIP interacting p29 from human samples.
PA1-31763 has been successfully used in Western blot applications.
The PA1-31763 immunogen is: Peptide with sequence AEIKQNLERGTAV, from C Terminus of the protein sequence according to NP_056299.
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.