PA1-7247 detects the Staphylococcus enterotoxin A. This antibody is not reactive with other Staphylococcus toxins.
PA1-7247 has been successfully used in ELISA procedures.
The PA1-7247 immunogen is purified toxin.
MA1-74347 recognises Staphylococcus aureus enterotoxin type A. Staphylococcus aureus produces several different types of enterotoxins (SE's). The SE's are short, soluble proteins that are highly stable and resistant to most proteolytic enzymes. They belong to a family of pyrogenic toxins and as such cause an increase in body temperature, immunosuppression and non-specific T-cell proliferation. These activities are referred to as superantigen activity and results from the interaction of SE's with T-cell antigen receptors and the major histocompatibility complex (MHC) on antigen presenting cells. In addition to their superantigen activity, SE's are also emetics (they cause vomiting). The SE's are important in food poisoning. Enterotoxin A is one of the SE's most commonly identified in clinical isolates. MA1-74347 may be used as a capture antibody in ELISA assays for S. aureus enterotoxin A in combination with MCA2848 as a detection agent.