    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DYKDDDDK Tag Antibodies
          OriGene
          DDK Monoclonal Antibody (OTI4C5 (formerly 4C5)), PE, TrueMAB™
          View all (123) DYKDDDDK Tag antibodies
          Tech Support
          Cite DDK Monoclonal Antibody (OTI4C5 (formerly 4C5)), PE, TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          DDK Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          DDK Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          DDK Antibody (TA150020) in Flow
          Normal CHO cells (black line) or CHO cells stably expressing FLAG®-tagged CB1 (red line) were fixed with 2% formaldehyde for 20 min at RT, stained with PE-labeled 4C5 anti-DDK antibody and analyzed by flow cytometry. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DDK Antibody in Flow Cytometry (Flow)
          Product Details
          TA150020
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          1:50
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI4C5 (formerly 4C5)
          Immunogen
          Anti-DDK monoclonal antibody is produced by immunizing mice with a synthetic peptide coupled to KLH
          Conjugate
          PE PE PE
          • Unconjugated
          • Agarose
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 594
          • HRP
          • Magnetic beads
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          TBS with 0.1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Synthetic peptide sequence is DYKDDDDK
          Target Information
          DYKDDDDK tag, also known as FLAG tag, is a short peptide sequence that is commonly used as a protein tag to facilitate protein purification, detection, and localization. The tag consists of eight amino acids: Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys. It is often fused to the N- or C-terminus of a protein of interest, allowing for easy identification and isolation of the protein using anti-FLAG antibodies. The DYKDDDDK tag is widely used in molecular biology and biochemistry research, and has been employed in various applications, such as protein-protein interaction studies, protein expression analysis, and protein localization studies. The FLAG tag structure has been optimized for compatibility with the proteins it is attached to, in that it is more hydrophilic than other common epitope tags and therefore less likely to denature or inactivate proteins to which it is appended. FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline