    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha Actinin 1 Antibodies
          OriGene
          alpha-actinin (Actinin alpha 1) Monoclonal Antibody (OTI7A4), TrueMAB™
          Advanced Verification
          2 References
          View all (22) alpha Actinin 1 antibodies
          Tech Support
          Cite alpha-actinin (Actinin alpha 1) Monoclonal Antibody (OTI7A4), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (11)
          • Advanced Verification (1)
          alpha-actinin (Actinin alpha 1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          alpha-actinin (Actinin alpha 1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 12
          alpha-actinin (Actinin alpha 1) Antibody (TA500072S) in ICC/IF
          Anti-ACTN1 mouse monoclonal antibody (TA500072) immunofluorescent staining of Hela cells transiently transfected by pCMV6-ENTRY ACTN1(RC204924). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          alpha-actinin (Actinin alpha 1) Antibody in Immunocytochemistry (ICC/IF)
          alpha-actinin (Actinin alpha 1) Antibody in Immunocytochemistry (ICC/IF)
          alpha-actinin (Actinin alpha 1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-actinin (Actinin alpha 1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-actinin (Actinin alpha 1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-actinin (Actinin alpha 1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-actinin (Actinin alpha 1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-actinin (Actinin alpha 1) Antibody in Western Blot (WB)
          alpha-actinin (Actinin alpha 1) Antibody in Western Blot (WB)
          alpha-actinin (Actinin alpha 1) Antibody in Western Blot (WB)
          alpha-actinin (Actinin alpha 1) Antibody in Western Blot (WB)
          alpha-actinin (Actinin alpha 1) Antibody
          alpha-actinin (Actinin alpha 1) Monoclonal Antibody (OTI7A4), TrueMAB™
          Product Details
          TA500072S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Miscellaneous PubMed (Misc)
          -
          View 2 publications 2 publications
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI7A4
          Immunogen
          Human recombinant protein fragment corresponding to amino acids 650-893 of human alpha-actinin produced in E.coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: actinin 1 smooth muscle; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; epididymis secretory sperm binding protein; F-actin cross-linking protein; FLJ40884; FLJ54432; Non-muscle alpha-actinin-1; unnamed protein product
          View more View less
          Gene Aliases: ACTN1; BDPLT15
          View more View less
          UniProt ID: (Human) P12814
          View more View less
          Entrez Gene ID: (Human) 87
          View more View less
          Function(s)
          transcription coactivator activity double-stranded RNA binding actin binding integrin binding calcium ion binding protein binding cytoskeletal protein binding vinculin binding identical protein binding protein homodimerization activity ion channel binding metal ion binding actin filament binding structural constituent of postsynapse
          Process(es)
          actin filament organization actin cytoskeleton organization platelet activation platelet formation platelet morphogenesis regulation of apoptotic process positive regulation of transcription, DNA-templated focal adhesion assembly anatomical structure formation involved in morphogenesis actin filament bundle assembly actin filament network formation muscle cell development platelet aggregation postsynapse organization
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline