The function of the PBLD protein remains unknown.
Protein Aliases: epididymis secretory protein Li 306; epididymis secretory sperm binding protein; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; Unknown protein 32 from 2D-page of liver tissue; unnamed protein product
Gene Aliases: HEL-S-306; MAWBP; MAWDBP; PBLD
UniProt ID: (Human) P30039
Entrez Gene ID: (Human) 64081