    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DEF8 Antibodies
          OriGene
          DEF8 Monoclonal Antibody (OTI2H4), TrueMAB™
          View all (8) DEF8 antibodies
          Tech Support
          Cite DEF8 Monoclonal Antibody (OTI2H4), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DEF8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DEF8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DEF8 Antibody (TA502670S) in WB
          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY DEF8 (RC224669, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-DEF8. Positive lysates LY403723 (100 µg) and LC403723 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DEF8 Antibody in Western Blot (WB)
          DEF8 Antibody in Flow Cytometry (Flow)
          DEF8 Monoclonal Antibody (OTI2H4), TrueMAB™
          Product Details
          TA502670S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Flow Cytometry (Flow)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI2H4
          Immunogen
          Full length human recombinant protein of human DEF8 produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.57 mg/mL
          Amount
          17.1 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Positively regulates lysosome peripheral distribution and ruffled border formation in osteoclasts. Involved in bone resorption. [UniProt]
          Bioinformatics
          Protein Aliases: DEF-8; Differentially expressed in FDCP 8 homolog; unnamed protein product
          View more View less
          Gene Aliases: DEF8
          View more View less
          UniProt ID: (Human) Q6ZN54
          View more View less
          Entrez Gene ID: (Human) 54849
          View more View less
          Function(s)
          protein binding zinc ion binding metal ion binding
          Process(es)
          lysosome localization positive regulation of bone resorption positive regulation of ruffle assembly
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline