    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PDF Antibodies
          OriGene
          PDF Monoclonal Antibody (OTI3C12), TrueMAB™
          View all (13) PDF antibodies
          Tech Support
          Cite PDF Monoclonal Antibody (OTI3C12), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          PDF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PDF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          PDF Antibody (TA502989S) in ICC/IF
          Anti-PDF mouse monoclonal antibody (TA502989) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY PDF(RC205788). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PDF Antibody in Immunocytochemistry (ICC/IF)
          PDF Antibody in Western Blot (WB)
          PDF Antibody in Western Blot (WB)
          PDF Antibody in Flow Cytometry (Flow)
          PDF Antibody in Flow Cytometry (Flow)
          PDF Monoclonal Antibody (OTI3C12), TrueMAB™
          Product Details
          TA502989S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3C12
          Immunogen
          Full length human recombinant protein of human PDF produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Removes the formyl group from the N-terminal Met of newly synthesized proteins.
          Bioinformatics
          Protein Aliases: huPD; Peptide deformylase, mitochondrial; peptide deformylase-like protein; Polypeptide deformylase
          View more View less
          Gene Aliases: PDF; PDF1A
          View more View less
          UniProt ID: (Human) Q9HBH1
          View more View less
          Entrez Gene ID: (Human) 64146
          View more View less
          Function(s)
          hydrolase activity peptide deformylase activity metal ion binding
          Process(es)
          translation positive regulation of cell proliferation peptidyl-methionine modification co-translational protein modification post-translational protein modification
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline