    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Alpha A Crystallin Antibodies
          OriGene
          CRYAA Monoclonal Antibody (OTI3B12), TrueMAB™
          View all (17) Alpha A Crystallin antibodies
          Tech Support
          Cite CRYAA Monoclonal Antibody (OTI3B12), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (9)
          CRYAA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CRYAA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          CRYAA Antibody (TA505577S) in ICC/IF
          Anti-CRYAA mouse monoclonal antibody (TA505577) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY CRYAA(RC216946). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CRYAA Antibody in Immunocytochemistry (ICC/IF)
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CRYAA Antibody in Western Blot (WB)
          CRYAA Monoclonal Antibody (OTI3B12), TrueMAB™
          Product Details
          TA505577S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:150
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3B12
          Immunogen
          Full length human recombinant protein of human CRYAA produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
          Bioinformatics
          Protein Aliases: Acry 1; Alpha-crystallin A chain; CRYA 1; CRYAA; Crystallin Alpha 1; crystallin, alpha-1; Heat shock protein beta-4; Heat shock protein family B member 4; HSPB 4; HspB4; human alphaA-crystallin (CRYA1)
          View more View less
          Gene Aliases: CRYA1; CRYAA; CRYAA2; CTRCT9; HSPB4
          View more View less
          UniProt ID: (Human) P02489
          View more View less
          Entrez Gene ID: (Human) 1409, (Human) 102724652
          View more View less
          Function(s)
          structural molecule activity structural constituent of eye lens protein binding identical protein binding metal ion binding unfolded protein binding
          Process(es)
          response to hypoxia lens development in camera-type eye lens morphogenesis in camera-type eye apoptotic process mitochondrion organization actin filament organization microtubule-based process tubulin complex assembly visual perception response to heat negative regulation of gene expression positive regulation of cell growth negative regulation of intracellular transport protein refolding response to hydrogen peroxide camera-type eye development negative regulation of apoptotic process embryonic camera-type eye morphogenesis protein stabilization apoptotic process involved in morphogenesis response to UV-A lens fiber cell morphogenesis
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline