    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • DPCD Antibodies
          OriGene
          DPCD Monoclonal Antibody (OTI4B9), TrueMAB™
          View all (7) DPCD antibodies
          Tech Support
          Cite DPCD Monoclonal Antibody (OTI4B9), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          DPCD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DPCD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          DPCD Antibody (TA505898S) in ICC/IF
          Anti-DPCD mouse monoclonal antibody (TA505898) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY DPCD(RC200890). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DPCD Antibody in Immunocytochemistry (ICC/IF)
          DPCD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DPCD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DPCD Antibody in Western Blot (WB)
          DPCD Monoclonal Antibody (OTI4B9), TrueMAB™
          Product Details
          TA505898S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:4,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:150
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI4B9
          Immunogen
          Full length human recombinant protein of human DPCD produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          May play a role in the formation or function of ciliated cells.
          Bioinformatics
          Protein Aliases: deleted in a mouse model of primary ciliary dyskinesia; Protein DPCD; RP11-529I10.4; RP11-529I104; unnamed protein product
          View more View less
          Gene Aliases: DPCD
          View more View less
          UniProt ID: (Human) Q9BVM2
          View more View less
          Entrez Gene ID: (Human) 25911
          View more View less
          Function(s)
          protein binding
          Process(es)
          epithelial cilium movement spermatogenesis determination of left/right symmetry ventricular system development lateral ventricle development third ventricle development sperm motility establishment of localization in cell left/right pattern formation
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline