CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Protein Aliases: CD105; CD105 antigen; Cell surface MJ7/18 antigen; circulating isoform of endoglin TGF-beta receptor type III; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); endoglin precursor; ORW; soluble endoglin; transmembrane glycoprotein; unnamed protein product
Gene Aliases: AI528660; AI662476; CD105; Edg; END; Endo; ENG; HHT1; ORW1; S-endoglin
UniProt ID: (Human) P17813, (Mouse) Q63961
Entrez Gene ID: (Dog) 609166, (Human) 2022, (Mouse) 13805, (Rat) 497010