    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • IL-1 alpha Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          OriGene
          IL1A Monoclonal Antibody (OTI3C3), TrueMAB™
          View all (57) IL-1 alpha antibodies
          Tech Support
          Cite IL1A Monoclonal Antibody (OTI3C3), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          IL1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          IL1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          IL1A Antibody (TA506853) in ICC/IF
          Anti-IL1A mouse monoclonal antibody (TA506853) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY IL1A(RC202084). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IL1A Antibody in Immunocytochemistry (ICC/IF)
          IL1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IL1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IL1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IL1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IL1A Antibody in Western Blot (WB)
          IL1A Monoclonal Antibody (OTI3C3), TrueMAB™
          Product Details
          TA506853
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:150
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3C3
          Immunogen
          Full length human recombinant protein of human IL1A produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline