          • Primary Antibodies ›
          • MUC6 Antibodies
          OriGene
          MUC6 Recombinant Rabbit Monoclonal Antibody (OTIR2F3), TrueRAB™
          View all (20) MUC6 antibodies
          Tech Support
          Cite MUC6 Recombinant Rabbit Monoclonal Antibody (OTIR2F3), TrueRAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MUC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          MUC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MUC6 Antibody (TA592120S) in IHC (P)
          Immunohistochemical staining of paraffin-embedded Human gastric tissue within the normal limits using anti-MUC6 rabbit monoclonal antibody. (Heat-induced epitope retrieval by 1mM EDTA in 10mM Tris buffer (pH8.0) at 120°C for 3 min, TA592120) (1:4000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MUC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC6 Recombinant Rabbit Monoclonal Antibody (OTIR2F3), TrueRAB™
          Product Details
          TA592120S
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:4,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          OTIR2F3
          Immunogen
          Synthetic peptide around the Ser1700 region of the human MUC6 (NP_005952) conjugated to KLH.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with 50% glycerol, 1% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
          Target Information
          Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.
          Bioinformatics
          Protein Aliases: gastric mucin; Gastric mucin-6; MUC-6; mucin 6; Mucin-6; unnamed protein product
          View more View less
          Gene Aliases: MUC-6; MUC6
          View more View less
          UniProt ID: (Human) Q6W4X9
          View more View less
          Entrez Gene ID: (Human) 4588
          View more View less
          Function(s)
          extracellular matrix structural constituent
          Process(es)
          maintenance of gastrointestinal epithelium
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline