    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TUBA8 Antibodies
          OriGene
          TUBA8 Monoclonal Antibody (OTI2E9), Biotin, TrueMAB™
          View all (22) TUBA8 antibodies
          Tech Support
          Cite TUBA8 Monoclonal Antibody (OTI2E9), Biotin, TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TUBA8 Antibody in Luminex (Luminex)
          Group 53 Created with Sketch.
          TUBA8 Antibody in Luminex (Luminex)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TUBA8 Antibody (TA700519) in Luminex
          TUBA8 Luminex with 2G6 Capture (TA600519) and 2E9 Detection (TA700519) Antibodies. Substrate used: Recombinant Human TUBA8 (TP311175) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TUBA8 Antibody in Luminex (Luminex)
          TUBA8 Monoclonal Antibody (OTI2E9), Biotin, TrueMAB™
          Product Details
          TA700519
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:250-1:1,000
          -
          Luminex (Luminex)
          Assay-Dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI2E9
          Immunogen
          Full length human recombinant protein of human TUBA8 produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 10mg/mL BSA, 50% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: Alpha-tubulin 8; TUBA8; tubulin alpha 8 trucated form; Tubulin alpha chain-like 2; Tubulin alpha-8 chain; unnamed protein product
          View more View less
          Gene Aliases: CDCBM8; MACTHC2; TUBA8; TUBAL2
          View more View less
          UniProt ID: (Human) Q9NY65
          View more View less
          Entrez Gene ID: (Human) 51807
          View more View less
          Function(s)
          nucleotide binding structural constituent of cytoskeleton protein binding GTP binding hydrolase activity metal ion binding
          Process(es)
          microtubule cytoskeleton organization mitotic cell cycle microtubule-based process spermatogenesis spermatid development
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline