          • Primary Antibodies ›
          • Complement Factor I Antibodies
          OriGene
          CFI Monoclonal Antibody (OTI7C9), TrueMAB™
          View all (57) Complement Factor I antibodies
          Tech Support
          Cite CFI Monoclonal Antibody (OTI7C9), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CFI Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CFI Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CFI Antibody (TA806003S) in WB
          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY CFI (RC216645, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-CFI. Positive lysates LY424865 (100 µg) and LC424865 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CFI Antibody in Western Blot (WB)
          CFI Monoclonal Antibody (OTI7C9), TrueMAB™
          Product Details
          TA806003S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-Dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI7C9
          Immunogen
          Human recombinant protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 µg/mL.
          Bioinformatics
          Protein Aliases: C3b-inactivator; C3B/C4B inactivator; CFI; complement component I; complement control protein factor I; Complement factor I; complement factor I heavy chain; factor i; Konglutinogen-activating factor; light chain of factor I; prepro-C3b/C4B inactivator; unnamed protein product
          View more View less
          Gene Aliases: AHUS3; ARMD13; C3b-INA; C3BINA; CFI; FI; IF; KAF
          View more View less
          UniProt ID: (Human) P05156
          View more View less
          Entrez Gene ID: (Human) 3426
          View more View less
          Function(s)
          serine-type endopeptidase activity protein binding peptidase activity serine-type peptidase activity hydrolase activity metal ion binding
          Process(es)
          immune system process proteolysis complement activation complement activation, classical pathway innate immune response
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline