FIGURE: 1 / 1
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems.
Protein Aliases: epididymis secretory sperm binding protein; Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; G alpha-15; G alpha-16; G protein; G-protein subunit alpha-16; GNA 15; GNA 16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); Guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16; heterotrimeric guanine nucleotide-binding protein 1L; unnamed protein product
Gene Aliases: GNA15; GNA16; HG1L
UniProt ID: (Human) P30679
Entrez Gene ID: (Human) 2769