    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • ZCCHC17 Antibodies
          OriGene
          ZCCHC17 Monoclonal Antibody (OTI9A7), TrueMAB™
          View all (4) ZCCHC17 antibodies
          Tech Support
          Cite ZCCHC17 Monoclonal Antibody (OTI9A7), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          ZCCHC17 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZCCHC17 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          ZCCHC17 Antibody (TA809551S) in WB
          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY ZCCHC17 (RC208652, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-ZCCH. Positive lysates LY413942 (100 µg) and LC413942 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZCCHC17 Antibody in Western Blot (WB)
          ZCCHC17 Monoclonal Antibody (OTI9A7), TrueMAB™
          Product Details
          TA809551S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI9A7
          Immunogen
          Full length human recombinant protein of human ZCCHC17 produced in E.coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          ZCCHC17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosolic large ribosomal subunit; nucleolus; nucleic acid binding; zinc ion binding.
          Bioinformatics
          Protein Aliases: hpNO40s; nucleolar protein 40; Nucleolar protein of 40 kDa; Pnn-interacting nucleolar protein; pNO40; PS1D protein; putative S1 RNA binding domain protein; Putative S1 RNA-binding domain protein; unnamed protein product; Zinc finger CCHC domain-containing protein 17; zinc finger, CCHC domain containing 17
          View more View less
          Gene Aliases: HSPC243; HSPC251; LDC4; pNO40; PS1D; ZCCHC17
          View more View less
          UniProt ID: (Human) Q9NP64
          View more View less
          Entrez Gene ID: (Human) 51538
          View more View less
          Function(s)
          nucleic acid binding RNA binding protein binding zinc ion binding identical protein binding metal ion binding
          Process(es)
          RNA stabilization
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline