    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RPAIN Antibodies
          OriGene
          RPAIN Monoclonal Antibody (OTI6A7), TrueMAB™
          View all (19) RPAIN antibodies
          Tech Support
          Cite RPAIN Monoclonal Antibody (OTI6A7), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          RPAIN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RPAIN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          RPAIN Antibody (TA810560S) in IHC (P)
          Immunohistochemical staining of paraffin-embedded human endometrium tissue within the normal limits using anti-RPAIN mouse monoclonal antibody. (Heat-induced epitope retrieval by 1mM EDTA in 10mM Tris buffer (pH8.5) at 120°C for 3 min, TA810560)(1:500). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RPAIN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RPAIN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RPAIN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RPAIN Antibody in Western Blot (WB)
          RPAIN Antibody in Western Blot (WB)
          RPAIN Monoclonal Antibody (OTI6A7), TrueMAB™
          Product Details
          TA810560S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI6A7
          Immunogen
          Full length human recombinant protein of human RPAIN produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
          Bioinformatics
          Protein Aliases: hRIP; importin; nuclear transporter; RAP interaction protein; RPA-interacting protein; unnamed protein product
          View more View less
          Gene Aliases: HRIP; RIP; RPAIN
          View more View less
          UniProt ID: (Human) Q86UA6
          View more View less
          Entrez Gene ID: (Human) 84268
          View more View less
          Function(s)
          protein binding zinc ion binding metal ion binding
          Process(es)
          protein import into nucleus response to UV
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline