    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • REV1 Antibodies
          OriGene
          REV1 Monoclonal Antibody (OTI1E12), TrueMAB™
          View all (9) REV1 antibodies
          Tech Support
          Cite REV1 Monoclonal Antibody (OTI1E12), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          REV1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          REV1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          REV1 Antibody (TA811575S) in WB
          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY REV1 (RC221742, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-REV1. (1. Positive lysates LY402538 (100 µg) and LC402538 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          REV1 Antibody in Western Blot (WB)
          REV1 Antibody in Western Blot (WB)
          REV1 Monoclonal Antibody (OTI1E12), TrueMAB™
          Product Details
          TA811575S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI1E12
          Immunogen
          Human recombinant protein fragment corresponding to amino acids 1079-1251 of human REV1 produced in E.coli.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a protein with similarity to the S. cerevisiae mutagenesis protein Rev1. The Rev1 proteins contain a BRCT domain, which is important in protein-protein interactions. A suggested role for the human Rev1-like protein is as a scaffold that recruits DNA polymerases involved in translesion synthesis (TLS) of damaged DNA. Two alternatively spliced transcript variants that encode different proteins have been found.
          Bioinformatics
          Protein Aliases: AIBP80; Alpha integrin-binding protein 80; DNA repair protein REV1; REV1 homolog; REV1, polymerase (DNA directed); REV1- like; Rev1-like terminal deoxycytidyl transferase
          View more View less
          Gene Aliases: AIBP80; REV1; REV1L
          View more View less
          UniProt ID: (Human) Q9UBZ9
          View more View less
          Entrez Gene ID: (Human) 51455, (Rat) 316344
          View more View less
          Function(s)
          damaged DNA binding protein binding deoxycytidyl transferase activity metal ion binding magnesium ion binding DNA-directed DNA polymerase activity DNA metabolism protein nucleic acid metabolism protein
          Process(es)
          DNA replication response to UV translesion synthesis error-prone translesion synthesis
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-659f68c6f4-jf685:80/100.66.76.102:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0