Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TAGAP Antibodies

          Abnova

          TAGAP Polyclonal Antibody, MaxPab™

          View all (8) TAGAP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TAGAP Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TAGAP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TAGAP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TAGAP Antibody (H00117289-D01P) in WB

          TAGAP MaxPab rabbit polyclonal antibody. Western Blot analysis of TAGAP expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TAGAP Antibody in Western Blot (WB)
          TAGAP Polyclonal Antibody, MaxPab™

          Product Details

          H00117289-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TAGAP (NP_473455.2, 1 a.a. approximately 731 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MKLRSSHNAS KTLNANNMET LIECQSEGDI KEHPLLASCE SEDSICQLIE VKKRKKVLSW PFLMRRLSPA SDFSGALETD LKASLFDQPL SIICGDSDTL PRPIQDILTI LCLKGPSTEG IFRRAANEKA RKELKEELNS GDAVDLERLP VHLLAVVFKD FLRSIPRKLL SSDLFEEWMG ALEMQDEEDR IEALKQVADK LPRPNLLLLK HLVYVLHLIS KNSEVNRMDS SNLAICIGPN MLTLENDQSL SFEAQKDLNN KVKTLVEFLI DNCFEIFGEN IPVHSSITSD DSLEHTDSSD VSTLQNDSAY DSNDPDVESN SSSGISSPSR QPQVPMATAA GLDSAGPQDA REVSPEPIVS TVARLKSSLA QPDRRYSEPS MPSSQECLES RVTNQTLTKS EGDFPVPRVG SRLESEEAED PFPEEVFPAV QGKTKRPVDL KIKNLAPGSV LPRALVLKAF SSSSLDASSD SSPVASPSSP KRNFFSRHQS FTTKTEKGKP SREIKKHSMS FTFAPHKKVL TKNLSAGSGK SQDFTRDHVP RGVRKESQLA GRIVQENGCE THNQTARGFC LRPHALSVDD VFQGADWERP GSPPSYEEAM QGPAARLVAS ESQTVGSMTV GSMRARMLEA HCLLPPLPPA HHVEDSRHRG SKEPLPGHGL SPLPERWKQS RTVHASGDSL GHVSGPGRPE LLPLRTVSES VQRNKRDCLV RRCSQPVFEA DQFQYAKESY I

          Target Information

          TAGAP (T-cell activation Rho GTPase-activating protein) is a 731 amino acid protein that contains one Rho-GAP domain and exists as 4 alternatively spliced isoforms. TAGAP may function as a GTPase-activating protein and may play important roles during T-cell activation. The gene that encodes TAGAP contains approximately 10,685 bases and maps to human chromosome 6q25.3. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, are also located on chromosome 6. In addition, Stickler syndrome, hydroxylase deficiency and maple syrup urine disease are associated with genes on chromosome 6, while a bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: T-cell activation GTPase-activating protein; T-cell activation Rho GTPase-activating protein; unnamed protein product

          View more View less

          Gene Aliases: ARHGAP47; FKSG15; IDDM21; TAGAP; TAGAP1

          View more View less

          UniProt ID: (Human) Q8N103

          View more View less

          Entrez Gene ID: (Human) 117289

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity GTPase activator activity protein binding
          Process(es)
          signal transduction regulation of Rho protein signal transduction regulation of small GTPase mediated signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline