Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total TBC1D3.
TBC1D3G acts as a GTPase activating protein for RAB5. Does not act on RAB4 or RAB11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Prostate cancer gene 17 protein; Protein TRE17-alpha; Rab GTPase-activating protein PRC17; TBC1 domain family member 3; TBC1 domain family member 3C; TBC1 domain family member 3C-like protein ENSP00000341742; TBC1 domain family member 3C/3D; unnamed protein product
Gene Aliases: PRC17; TBC1D3; TBC1D3A; TBC1D3C; TBC1D3D
UniProt ID: (Human) Q8IZP1
Entrez Gene ID: (Human) 414060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support