Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAEARKRRE LLPLIYHHLL RAGYVRAARE VKEQSGQKCF LAQPVTLLDI YTHWQQTSEL GRKRKAEEDA ALQAKKTRVS DPISTSESSE EEEEAEAETA KATPRLASTN SSVLGADLPS SMKEKAKAET EKAGKTGNSM PHPATGKTVA NLLSGKSPRK SAEPSANTTL VSETEEEGSV PAFGAAAKPG MVSAGQADSS SEDTSSSSDE TDVEGKPSVK PAQVKASSVS TKESPARKAA PAPGKVGDVT PQVKGGALPP AKRAKKPEEE SESSEEGSES EEEAPAGTRS QVKASEKILQ VRAASAPAKG TPGKGATPAP PGKAGAVASQ TKAGKPEEDS ESSSEESSDS E
Treacle is a serine/alanine-rich 1,411 amino acid nuclear phosphoprotein that interacts with upstream binding factor (UBF) and affects transcription of the ribosomal DNA gene. Treacle is also involved in early embryonic development, particularly in the craniofacial complex, and may play a role in nucleolarcytoplasmic transport. The Treacle protein contains three domains with unique N- and C-termini and a large central repeat domain. Mutations in TCOF1, the gene that encodes for Treacle, cause Treacher Collins-Franceschetti syndrome (TCS), a disorder characterized by defects in craniofacial development. Symptoms of TCS include conductive hearing loss, hypoplasia of the mandible and maxilla, downward sloping palpebral fissures and cleft palate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nucleolar phosphoprotein; nucleolar trafficking phosphoprotein; putative nucleolar phosphoprotein; Treacher Collins Franceschetti syndrome 1, homolog; Treacher Collins syndrome; Treacher Collins syndrome protein; treacher Collins syndrome protein homolog; Treacher Collins-Franceschetti syndrome 1; unnamed protein product
Gene Aliases: MFD1; TCS; TCS1; treacle
UniProt ID: (Human) Q13428, (Mouse) O08784
Entrez Gene ID: (Human) 6949, (Mouse) 21453
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support