Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MLRSCAARL RTLGALCRPP VGRRLPGSEP RPELRSFSSE EVILKDCSVP NPSWNKDLRL LFDQFMKKCE DGSWKRLPSY KRTPTEWIQD FKTHFLDPKL MKEEQMSQAQ LFTRSFDDGL GFEYVMFYND IEKRMVCLFQ GGPYLEGPPG FIHGGAIATM IDATVGMCAM MAGGIVMTAN LNINYKRPIP LCSVVMINSQ LDKVEGRKFF VSCNVQSVDE KTLYSEATSL FIKLNPAKSL T (1-240 aa encoded by BC065277)
THEM4, also known as CTMP, binds specifically to the carboxy-terminal regulatory domain of PKB/Akt at the plasma membrane and acts as a negative regulator, reversing the phenotype of v-Akt-transformed cells. Hypermethylation of the THEM4 promoter and transcriptional downregulation of the gene has been reported in multiple glioblastomas, suggesting that epigenetic regulation of THEM4 may play a role in the progression of this cancer. Bioinformatic analysis, confirmed by in vitro testing, indicates that THEM4 is a broad-range, high activity acyl-CoA thioesterase. Recent reports have also indicated that TMEM4 is a mitochondrial protein whose overexpression is associated with an increase in mitochondrial membrane depolarization and caspase-3 and PARP cleavage, suggesting that THEM4 is involved in the apoptotic program.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA thioesterase THEM4; C-terminal modulator protein; carboxyl-terminal modulator protein; unnamed protein product
Gene Aliases: 2700077M13Rik; 4921507I02Rik; CTMP; RGD1304723
UniProt ID: (Human) Q5T1C6, (Mouse) Q3UUI3, (Rat) Q566R0
Entrez Gene ID: (Human) 117145, (Mouse) 75778, (Rat) 361992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support