Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TIMELESS Antibodies

          Proteintech

          TIMELESS Polyclonal Antibody

          4 References
          View all (17) TIMELESS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TIMELESS Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (18)
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIMELESS Antibody (14421-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human placenta using 14421-1-AP (TIMELESS antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIMELESS Antibody in Western Blot (WB)
          TIMELESS Antibody in Western Blot (WB)
          TIMELESS Antibody in Western Blot (WB)
          TIMELESS Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TIMELESS Polyclonal Antibody

          Product Details

          14421-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TIMELESS Fusion Protein Ag5767 (869-1208 aa encoded by BC050557)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: QMGLADSVK DFQRKGTHIV LWTGDQELEL QRLFEEFRDS DDVLGHIMKN ITAKRSRARI VDKLLALGLV AERRELYKKR QKKLASSILP NGAESLKDFC QEDLEEEENL PEEDSEEEEE GGSEAEQVQG SLVLSNENLG QSLHQEGFSI LLLWLQNCLI RAADDREEDG CSQAVPLVPL TEENEEAMEN EQFQQLLRKL GVRPPASGQE TFWRIPAKLS PTQLRRAAAS LSQPEEEQKL QPELQPKVPG EQGSDEEHCK EHRAQALRAL LLAHKKKAGL ASPEEEDAVG KEPLKAAPKK RQLLDSDEEQ EEDEGRNRAP ELGAPGIQKK KRYQIEDDED D (869-1208 aa encoded by BC050557)

          Target Information

          The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hTIM; mTIM; timeless; timeless circadian clock 1; timeless homolog; Tof1 homolog; unnamed protein product

          View more View less

          Gene Aliases: Debt69; FASPS4; hTIM; TIM; TIM1

          View more View less

          UniProt ID: (Human) Q9UNS1, (Mouse) Q9R1X4

          View more View less

          Entrez Gene ID: (Human) 8914, (Mouse) 21853

          View more View less

          Function(s)
          DNA binding protein binding enzyme activator activity identical protein binding
          Process(es)
          DNA replication checkpoint negative regulation of transcription from RNA polymerase II promoter branching involved in ureteric bud morphogenesis kidney development morphogenesis of an epithelium DNA repair cellular response to DNA damage stimulus circadian rhythm lung development replication fork processing intra-S DNA damage checkpoint regulation of circadian rhythm positive regulation of circadian rhythm replication fork arrest cell cycle phase transition negative regulation of transcription, DNA-templated branching morphogenesis of an epithelial tube cellular response to hydroxyurea cellular response to cisplatin cellular response to bleomycin positive regulation of double-strand break repair via homologous recombination positive regulation of double-strand break repair detection of abiotic stimulus regulation of cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-lz9xs:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0