Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TIMELESS Antibodies

          Abnova

          TIMELESS Polyclonal Antibody, MaxPab™

          View all (16) TIMELESS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TIMELESS Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          TIMELESS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TIMELESS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIMELESS Antibody (H00008914-B01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to TIMELESS on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIMELESS Antibody in Immunocytochemistry (ICC/IF)
          TIMELESS Antibody in Western Blot (WB)
          TIMELESS Polyclonal Antibody, MaxPab™

          Product Details

          H00008914-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TIMELESS (NP_003911.1, 1 a.a. approximately 1208 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MDLHMMNCEL LATCSALGYL EGDTYHKEPD CLESVKDLIR YLRHEDETRD VRQQLGAAQI LQSDLLPILT QHHQDKPLFD AVIRLMVNLT QPALLCFGNL PKEPSFRHHF LQVLTYLQAY KEAFASEKAF GVLSETLYEL LQLGWEERQE EDNLLIERIL LLVRNILHVP ADLDQEKKID DDASAHDQLL WAIHLSGLDD LLLFLASSSA EEQWSLHVLE IVSLMFRDQN PEQLAGVGQG RLAQERSADF AELEVLRQRE MAEKKTRALQ RGNRHSRFGG SYIVQGLKSI GERDLIFHKG LHNLRNYSSD LGKQPKKVPK RRQAARELSI QRRSALNVRL FLRDFCSEFL ENCYNRLMGS VKDHLLREKA QQHDETYYMW ALAFFMAFNR AASFRPGLVS ETLSVRTFHF IEQNLTNYYE MMLTDRKEAA SWARRMHLAL KAYQELLATV NEMDLSPDEA VRESSRIIKN NIFYVMEYRE LFLALFRKFD ERCQPRSFLR DLVETTHLFL KMLERFCRSR GNLVVQNKQK KRRKKKKKVL DQAIVSGNVP SSPEEVEAVW PALAEQLQCC AQNSELSMDS VVPFDAASEV PVEEQRAEAM VRIQDCLLAG QAPQALTLLR SAREVWPEGD VFGSQDISPE EEIQLLKQIL SAPLPRQQGP EERGAEEEEE EEEEEEEELQ VVQVSEKEFN FLDYLKRFAC STVVRAYVLL LRSYQQNSAH TNHCIVKMLH RLAHDLKMEA LLFQLSVFCL FNRLLSDPAA GAYKELVTFA KYILGKFFAL AAVNQKAFVE LLFWKNTAVV REMTEGYGSL DDRSSSRRAP TWSPEEEAHL QELYLANKDV EGQDVVEAIL AHLNTVPRTR KQIIHHLVQM GLADSVKDFQ RKGTHIVLWT GDQELELQRL FEEFRDSDDV LGHIMKNITA KRSRARIVDK LLALGLVAER RELYKKRQKK LASSILPNGA ESLKDFCQED LEEEENLPEE DSEEEEEGGS EAEQVQGSLV LSNENLGQSL HQEGFSIPLL WLQNCLIRAA DDREEDGCSQ AVPLVPLTEE NEEAMENEQF QQLLRKLGVR PPASGQETFW RIPAKLSPTQ LRRAAASLSQ PEEEQKLQPE LQPKVPGEQG SDEEHCKEHR AQALRALLLA HKKKAGLASP EEEDAVGKEP LKAAPKKRQL LDSDEEQEED EGRNRAPELG APGIQKKKRY QIEDDEDD

          Target Information

          The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hTIM; Protein timeless homolog; timeless circadian clock 1; timeless homolog; Tof1 homolog; unnamed protein product

          View more View less

          Gene Aliases: FASPS4; hTIM; TIM; TIM1; TIMELESS; TIMELESS1

          View more View less

          UniProt ID: (Human) Q9UNS1

          View more View less

          Entrez Gene ID: (Human) 8914

          View more View less

          Function(s)
          DNA binding protein binding enzyme activator activity identical protein binding
          Process(es)
          DNA replication checkpoint morphogenesis of an epithelium DNA repair cellular response to DNA damage stimulus circadian rhythm detection of abiotic stimulus lung development replication fork processing regulation of cell proliferation regulation of circadian rhythm replication fork arrest cell cycle phase transition negative regulation of transcription, DNA-templated rhythmic process branching morphogenesis of an epithelial tube cell division cellular response to hydroxyurea cellular response to cisplatin cellular response to bleomycin positive regulation of double-strand break repair via homologous recombination positive regulation of double-strand break repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-xwh6v:80/100.66.74.186:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline