Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • TIN2 Antibodies

          Abnova

          TINF2 Polyclonal Antibody, MaxPab™

          View all (14) TIN2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TINF2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TINF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TINF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TINF2 Antibody (H00026277-B01P) in WB

          Western Blot analysis of TINF2 expression in transfected 293T cell line by TINF2 MaxPab polyclonal antibody. Lane 1: TINF2 transfected lysate(49.61 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TINF2 Antibody in Western Blot (WB)
          TINF2 Polyclonal Antibody, MaxPab™

          Product Details

          H00026277-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TINF2 (AAH05030.1, 1 a.a. approximately 451 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MATPLVAGPA ALRFAAAASW QVVRGRCVEH FPRVLEFLRS LRAVAPGLVR YRHHERLCMG LKAKVVVELI LQGRPWAQVL KALNHHFPES GPIVRDPKAT KQDLRKILEA QETFYQQVKQ LSEAPVDLAS KLQELEQEYG EPFLAAMEKL LFEYLCQLEK ALPTPQAQQL QDVLSWMQPG VSITSSLAWR QYGVDMGWLL PECSVTDSVN LAEPMEQNPP QQQRLALHNP LPKAKPGTHL PQGPSSRTHP EPLAGRHFNL APLGRRRVQS QWASTRGGHK ERPTVMLFPF RNLGSPTQVI SKPESKEEHA IYTADLAMGT RAASTGKSKS PCQTLGGRAL KENPVDLPAT EQKENCLDCY MDPLRLSLLP PRARKPVCPP SLCSSVITIG DLVLDSDEEE NGQGEGKESL ENYQKTKFDT LIPTLCEYLP PSGHGAIPVS SCDCRDSSRP L

          Target Information

          TRF1 is a negative regulator of telomere length maintenance. Recently two proteins, tankyrase and Tin2, which bind to mammalian TRF1, have been identified. Tankyrase is a protein with homology to ankyrins as well as to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase localizes to telomeres by binding to the telomeric repeat binding factor 1 (TRF1) through its ankyrin repeats. Tankyrase exhibits PARP activity functioning as acceptors for adenosine diphosphate (ADP)-ribosylation. Since ADP-ribosylation of TRF1 diminishes its ability to bind to telomeric DNA, this suggests that telomere function in human cells is regulated by poly (ADP)-ribosylation. Both the cell cycle and TRF1 may regulate the subcellular localization of tankyrase. The cDNA coding for TIN2 protein was isolated as interacting partner of TRF1 from a yeast two-hybrid cDNA library screening. Tin2 localizes to telomeres and is essential for proper regulation of telomere length. TIN2, like TRF1, is widely and constitutively expressed, suggesting that these proteins act together to counterbalance telomere elongation by telomerase. However, endogenous TIN2, like endogenousTRF1, is not highly expressed, and therefore may be difficult to visualize by immunostaining in certain cell systems.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: TERF1 (TRF1)-interacting nuclear factor 2; TERF1-interacting nuclear factor 2; tin; Tin-2; TRF1-interacting nuclear factor 2; telomere-associated protein; TRF1-interacting nuclear protein 2; unnamed protein product

          View more View less

          Gene Aliases: DKCA3; DKCA5; TIN2; TINF2

          View more View less

          UniProt ID: (Human) Q9BSI4

          View more View less

          Entrez Gene ID: (Human) 26277

          View more View less

          Function(s)
          DNA binding protein binding telomeric DNA binding
          Process(es)
          telomere capping telomere assembly positive regulation of telomere maintenance negative regulation of telomere maintenance via telomerase negative regulation of epithelial cell proliferation protein localization to chromosome, telomeric region regulation of telomere maintenance via telomere lengthening
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline