Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TLE Pan Antibodies

          Invitrogen

          TLE Pan Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TLE Pan Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TLE Pan Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TLE Pan Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TLE Pan Antibody (PA5-17241) in WB

          Western blot analysis of TLE1/2/3/4 in cell extracts from LN18 and SW620 cells using TLE1/2/3/4 polyclonal antibody (Product # PA5-17241). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TLE Pan Antibody in Western Blot (WB)
          TLE Pan Polyclonal Antibody

          Product Details

          PA5-17241

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Fruit fly, Human, Mouse, Non-human primate

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues surrounding Asp760 of human TLE1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          261 µg/mL

          Amount

          26.1 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10982642

          Product Specific Information

          It is not recommended to aliquot this antibody.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: B lymphocyte gene 1; E(Sp1) homolog; enhancer of split groucho 3; enhancer of split groucho 4; Enhancer of split groucho-like protein 1; Enhancer of split groucho-like protein 2; Enhancer of split groucho-like protein 3; ESG; ESG-1; ESG-2; ESG-3; ESG-4; ESG1; ESG2; ESG3; Grg-1; Grg-3; Grg-4; groucho-related protein; Groucho-related protein 1; Groucho-related protein 4; transducin like enhancer of split 1; transducin like enhancer of split 2; transducin like enhancer of split 3; transducin like enhancer of split 4; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); Transducin-like enhancer protein 1; Transducin-like enhancer protein 2; Transducin-like enhancer protein 3; Transducin-like enhancer protein 4; unnamed protein product

          View more View less

          Gene Aliases: 2610103N05Rik; 5730411M05Rik; AA792082; BCE-1; BCE1; C230057C06Rik; E(spI); E(spl); ESG; ESG1; ESG2; ESG3; ESG4; ESTM13; ESTM14; Grg-1; Grg-4; GRG1; GRG2; GRG3; Grg3a; Grg3b; GRG4; HsT18976; KIAA1261; KIAA1547; mKIAA1547; mKIAA4188; TLE-1; TLE1; TLE2; TLE3; TLE4; Tle4l; X83332; X83333

          View more View less

          UniProt ID: (Human) Q04724, (Human) Q04725, (Human) Q04726, (Human) Q04727, (Mouse) Q62440, (Mouse) Q9WVB2, (Mouse) Q08122, (Mouse) Q62441

          View more View less

          Entrez Gene ID: (Human) 7088, (Human) 7089, (Human) 7090, (Human) 7091, (Mouse) 21885, (Mouse) 21886, (Mouse) 21887, (Mouse) 21888

          View more View less

          Function(s)
          transcription corepressor activity protein binding identical protein binding DNA-binding transcription factor binding chromatin binding RNA polymerase II transcription corepressor activity transcription factor binding transcription cofactor
          Process(es)
          regulation of transcription, DNA-templated signal transduction organ morphogenesis positive regulation of gene expression Wnt signaling pathway negative regulation of Wnt signaling pathway negative regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of transcription, DNA-templated negative regulation of canonical Wnt signaling pathway negative regulation of anoikis negative regulation of cold-induced thermogenesis negative regulation of transcription from RNA polymerase II promoter cellular response to leukemia inhibitory factor transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter multicellular organism development spermatogenesis cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0