Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TNNI3K Antibodies

          Abnova

          TNNI3K Polyclonal Antibody, MaxPab™

          View all (6) TNNI3K antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TNNI3K Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TNNI3K Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TNNI3K Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TNNI3K Antibody (H00051086-D01P) in WB

          TNNI3K MaxPab rabbit polyclonal antibody. Western Blot analysis of TNNI3K expression in A-431. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TNNI3K Antibody in Western Blot (WB)

          Product Details

          H00051086-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TNNI3K (AAH32865.1, 1 a.a. approximately 843 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAARDPPEV SLREATQRKL RRFSELRGKL VARGEFWDIV AITAADEKQE LAYNQQLSEK LKRKELPLGV QYHVFVDPAG AKIGNGGSTL CALQCLEKLY GDKWNSFTIL LIHSDEWKKK VSESYVITIE RLEDDLQIKE KELTELRNIF GSDEAFSKVN LNYRTENGLS LLHLCCICGG KKSHIRTLML KGLRPSRLTR NGFTALHLAV YKDNAELITS LLHSGADVQQ VGYGGLTALH IATIAGHLEA ADVLLQHGAN VNIQDAVFFT PLHIAAYYGH EQVTRLLLKF GADVNVSGEV GDRPLHLASA KGFLNIAKLL MEEGSKADVN AQDNEDHVPL HFCSRFGHHD IVKYLLQSDL EVQPHVVNIY GDTPLHLACY NGKFEVAKEI IQISGTESLT KENIFSETAF HSACTYGKSI DLVKFLLDQN VININHQGRD GHTGLHSACY HGHIRLVQFL LDNGADMNLV ACDPSRSSGE KDEQTCLMWA YEKGHDAIVT LLKHYKRPQD ELPCNEYSQP GGDGSYVSVP SPLGKIKSMT KEKADILLLR AGLPSHFHLQ LSEIEFHEII GSGSFGKVYK GRCRNKIVAI KRYRANTYCS KSDVDMFCRE VSILCQLNHP CVIQFVGACL NDPSQFAIVT QYISGGSLFS LLHEQKRILD LQSKLIIAVD VAKGMEYLHN LTQPIIHRDL NSHNILLYED GHAVVADFGE SRFLQSLDED NMTKQPGNLL WMAPEVFTQC TRYTIKADVF SYALCLWEIL TGEIPFAHLK PAAAAADMAY HHIRPPIGYS IPKPISSLLI RGWNACPEAK SRPSHYPVSS VYTETLKKKN EDRFGMWIEY LRR

          Target Information

          TNNI3K may play a role in cardiac physiology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cardiac ankyrin repeat kinase; Cardiac troponin I-interacting kinase; CARK; Serine/threonine-protein kinase TNNI3K; TNNI3-interacting kinase; tnni3k

          View more View less

          Gene Aliases: CARK; CCDD; D830019J24Rik; TNNI3K

          View more View less

          UniProt ID: (Human) Q59H18, (Mouse) Q5GIG6

          View more View less

          Entrez Gene ID: (Human) 51086, (Mouse) 435766

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity metal ion binding protein serine kinase activity protein C-terminus binding transferase activity, transferring phosphorus-containing groups troponin I binding non-receptor serine/threonine protein kinase
          Process(es)
          regulation of heart rate protein phosphorylation peptidyl-serine autophosphorylation regulation of cardiac muscle contraction bundle of His cell to Purkinje myocyte communication regulation of cardiac conduction phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline