Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TNXB Antibodies

          Abnova

          TNXB Polyclonal Antibody, MaxPab™

          View all (10) TNXB antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TNXB Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TNXB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TNXB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TNXB Antibody (H00007148-B01P) in WB

          Western Blot analysis of TNXB expression in transfected 293T cell line (H00007148-T01) by TNXB MaxPab polyclonal antibody.

          Lane 1: TNXB transfected lysate(74.03 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TNXB Antibody in Western Blot (WB)
          TNXB Polyclonal Antibody, MaxPab™

          Product Details

          H00007148-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TNXB (AAH33740.1, 1 a.a. approximately 673 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MRLSWSVAQG PFDSFVVQYE DTNGQPQALL VDGDQSKILI SGLEPSTPYR FLLYGLHEGK RLGPLSAEGT TGLAPAGQTS EESRPRLSQL SVTDVTTSSL RLNWEAPPGA FDSFLLRFGV PSPSTLEPHP RPLLQRELMV PGTRHSAVLR DLRSGTLYSL TLYGLRGPHK ADSIQGTART LSPVLESPRD LQFSEIRETS AKVNWMPPPS RADSFKVSYL LADGGEPQSV QVDGQARTQK LQGLIPGARY EVTVVSVRGF EESEPLTGFL TTVPDGPTQL RALNLTEGFA VLHWKPPQNP VDTYDVQVTA PGAPPLQAET PGSAVDYPLH DLVLHTNYTA TVRGLRGPNL TSPASITFTT GLEAPRDLEA KEVTPRTALL TWTEPPVRPA GYLLSFHTPG GQNQEILLPG GITSHQLLGL FPSTSYNARL QAMWGQSLLP PVSTSFTTGG LRIPFPRDCG EEMQNGAGAS RTSTIFLNGN RERPLNVFCD METDGGGWLV FQRRMDGQTD FWRDWEDYAH GFGNISGEFW LGNEALHSLT QAGDYSMRVD LRAGDEAVFA QYDSFHVDSA AEYYRLHLEG YHGTAGDSMS YHSGSVFSAR DRDPNSLLIS CAVSYRGAWW YRNCHYANLN GLYGSTVDHQ GVSWYHWKGF EFSVPFTEMK LRPRNFRSPA GGG

          Target Information

          Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GIG45; growth-inhibiting protein 45; Hexabrachion-like protein; tenascin XB1; tenascin XB2; Tenascin-X; TN-X; TNXB2tenascin XB1; TNXBSTNXTENX; unidentified protein; XB-S; XBTN-X

          View more View less

          Gene Aliases: EDS3; EDSCLL; EDSCLL1; HXBL; TENX; TN-X; TNX; TNXB; TNXB1; TNXB2; TNXBS; VUR8; XB; XBS

          View more View less

          UniProt ID: (Human) P22105

          View more View less

          Entrez Gene ID: (Human) 7148

          View more View less

          Function(s)
          integrin binding extracellular matrix structural constituent protein binding collagen binding heparin binding collagen fibril binding
          Process(es)
          lipid metabolic process fatty acid metabolic process triglyceride metabolic process cell adhesion cell-matrix adhesion positive regulation of cell proliferation positive regulation of epithelial to mesenchymal transition actin cytoskeleton organization regulation of cell adhesion extracellular matrix organization collagen fibril organization regulation of cell migration neuron projection development collagen metabolic process regulation of cell differentiation regulation of JNK cascade elastic fiber assembly cell-cell adhesion positive regulation of vascular endothelial growth factor signaling pathway positive regulation of collagen fibril organization positive regulation of cell fate determination
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline